Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 16200404-16200651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT15
Entrez Gene ID
117248
Gene Name
polypeptide N-acetylgalactosaminyltransferase 15
Gene Aliases
GALNACT15, GALNT13, GALNT7, GALNTL2, PIH5, pp-GalNAc-T15
UniGene
Hs.411308
Gene Chromosome Location
Chr.3: 16174322-16245190 on Build GRCh38
Transcripts
NM_001319051.1 NM_054110.4 XM_005264852.4 XM_017005682.1 XM_017005683.1 XM_017005684.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3940372 c.735T>A p.Y245* Chr.3: 16200647-16200647 on Build GRCh38
222781 c.730G>A p.E244K Chr.3: 16200642-16200642 on Build GRCh38
3202529 c.707G>A p.G236E Chr.3: 16200619-16200619 on Build GRCh38
7207797 c.714C>G p.L238L Chr.3: 16200626-16200626 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs541620439 Chr.3: 16200509-16200509 on Build GRCh38
rs371302412 Chr.3: 16200604-16200604 on Build GRCh38
rs769735098 Chr.3: 16200616-16200616 on Build GRCh38
rs777455659 Chr.3: 16200545-16200545 on Build GRCh38
rs1036483433 Chr.3: 16200445-16200445 on Build GRCh38
rs200118171 Chr.3: 16200619-16200619 on Build GRCh38
rs557520950 Chr.3: 16200623-16200623 on Build GRCh38
rs1011794669 Chr.3: 16200630-16200630 on Build GRCh38
rs563180953 Chr.3: 16200498-16200498 on Build GRCh38
rs757260501 Chr.3: 16200588-16200588 on Build GRCh38
rs773048639 Chr.3: 16200618-16200618 on Build GRCh38
rs534893826 Chr.3: 16200597-16200599 on Build GRCh38
rs879541021 Chr.3: 16200602-16200602 on Build GRCh38
rs944355966 Chr.3: 16200629-16200629 on Build GRCh38
rs549532849 Chr.3: 16200476-16200476 on Build GRCh38
rs746085597 Chr.3: 16200599-16200599 on Build GRCh38
rs3796301 Chr.3: 16200594-16200594 on Build GRCh38
rs746474453 Chr.3: 16200546-16200546 on Build GRCh38
rs930276992 Chr.3: 16200494-16200494 on Build GRCh38
rs200995755 Chr.3: 16200606-16200606 on Build GRCh38
rs1009409019 Chr.3: 16200520-16200520 on Build GRCh38
rs576634372 Chr.3: 16200510-16200510 on Build GRCh38
rs1020899309 Chr.3: 16200480-16200480 on Build GRCh38
rs762247025 Chr.3: 16200596-16200596 on Build GRCh38
rs1050108724 Chr.3: 16200497-16200497 on Build GRCh38
rs957656983 Chr.3: 16200472-16200472 on Build GRCh38
rs958732419 Chr.3: 16200569-16200569 on Build GRCh38
rs759127078 Chr.3: 16200628-16200628 on Build GRCh38
rs377760282 Chr.3: 16200570-16200570 on Build GRCh38
rs534853378 Chr.3: 16200621-16200621 on Build GRCh38
rs371247733 Chr.3: 16200573-16200573 on Build GRCh38
rs79184660 Chr.3: 16200453-16200453 on Build GRCh38
rs962488322 Chr.3: 16200574-16200574 on Build GRCh38
rs114904409 Chr.3: 16200578-16200578 on Build GRCh38
rs546028680 Chr.3: 16200513-16200513 on Build GRCh38
rs747965189 Chr.3: 16200612-16200612 on Build GRCh38
rs533662265 Chr.3: 16200547-16200547 on Build GRCh38
rs966455583 Chr.3: 16200496-16200496 on Build GRCh38
rs753068969 Chr.3: 16200571-16200571 on Build GRCh38
rs187108016 Chr.3: 16200564-16200564 on Build GRCh38
rs756546291 Chr.3: 16200572-16200572 on Build GRCh38

View Full Product Details