Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65018413-65018672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL2
Entrez Gene ID
402
Gene Name
ADP ribosylation factor like GTPase 2
Gene Aliases
ARFL2
UniGene
Hs.502836
Gene Chromosome Location
Chr.11: 65014113-65022185 on Build GRCh38
Transcripts
NM_001199745.1 NM_001667.3 NR_037650.1 NR_106939.1

Gene Symbol
ARL2-SNX15
Entrez Gene ID
100528018
Gene Name
ARL2-SNX15 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.80132
Gene Chromosome Location
Chr.11: 65014113-65040572 on Build GRCh38
Transcripts
NM_001199745.1 NM_001667.3 NR_037650.1 NR_106939.1

Gene Symbol
MIR6879
Entrez Gene ID
102466756
Gene Name
microRNA 6879
Gene Aliases
hsa-mir-6879
UniGene
Gene Chromosome Location
Chr.11: 65018505-65018570 on Build GRCh38
Transcripts
NM_001199745.1 NM_001667.3 NR_037650.1 NR_106939.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
397433 c.209A>G p.Q70R Chr.11: 65018603-65018603 on Build GRCh38
3810004 c.221G>T p.R74L Chr.11: 65018615-65018615 on Build GRCh38
6194326 c.119A>G p.D40G Chr.11: 65018417-65018417 on Build GRCh38
3726594 c.127A>G p.T43A Chr.11: 65018425-65018425 on Build GRCh38
1283501 c.248G>A p.S83N Chr.11: 65018642-65018642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937482758 Chr.11: 65018435-65018435 on Build GRCh38
rs543741844 Chr.11: 65018601-65018601 on Build GRCh38
rs761946079 Chr.11: 65018470-65018470 on Build GRCh38
rs770092004 Chr.11: 65018452-65018452 on Build GRCh38
rs761010941 Chr.11: 65018485-65018485 on Build GRCh38
rs570221663 Chr.11: 65018558-65018558 on Build GRCh38
rs750050216 Chr.11: 65018539-65018539 on Build GRCh38
rs751493254 Chr.11: 65018514-65018514 on Build GRCh38
rs773435992 Chr.11: 65018483-65018483 on Build GRCh38
rs144161068 Chr.11: 65018460-65018460 on Build GRCh38
rs773171394 Chr.11: 65018627-65018627 on Build GRCh38
rs745559338 Chr.11: 65018451-65018451 on Build GRCh38
rs774777357 Chr.11: 65018469-65018469 on Build GRCh38
rs779442670 Chr.11: 65018607-65018607 on Build GRCh38
rs780425731 Chr.11: 65018453-65018453 on Build GRCh38
rs755763132 Chr.11: 65018553-65018553 on Build GRCh38
rs201379160 Chr.11: 65018555-65018555 on Build GRCh38
rs748651201 Chr.11: 65018615-65018615 on Build GRCh38
rs772153074 Chr.11: 65018621-65018621 on Build GRCh38
rs764092292 Chr.11: 65018513-65018513 on Build GRCh38
rs906537666 Chr.11: 65018561-65018561 on Build GRCh38
rs771266504 Chr.11: 65018646-65018646 on Build GRCh38
rs777063183 Chr.11: 65018647-65018647 on Build GRCh38
rs74814065 Chr.11: 65018557-65018557 on Build GRCh38
rs199961468 Chr.11: 65018471-65018471 on Build GRCh38
rs752554416 Chr.11: 65018487-65018487 on Build GRCh38
rs769161323 Chr.11: 65018462-65018462 on Build GRCh38
rs762903453 Chr.11: 65018490-65018490 on Build GRCh38
rs765057682 Chr.11: 65018486-65018486 on Build GRCh38
rs202163474 Chr.11: 65018523-65018523 on Build GRCh38
rs760922486 Chr.11: 65018641-65018641 on Build GRCh38
rs780586325 Chr.11: 65018534-65018534 on Build GRCh38
rs749724483 Chr.11: 65018457-65018457 on Build GRCh38
rs369088336 Chr.11: 65018565-65018565 on Build GRCh38
rs1056004482 Chr.11: 65018439-65018439 on Build GRCh38

View Full Product Details