Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 231258906-231259177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC9
Entrez Gene ID
80210
Gene Name
armadillo repeat containing 9
Gene Aliases
ARM, KU-MEL-1, NS21
UniGene
Hs.162411
Gene Chromosome Location
Chr.2: 231198546-231394991 on Build GRCh38
Transcripts
NM_001271466.2 NM_001291656.1 NM_025139.5 XM_011511905.1 XM_011511906.1 XM_011511907.1 XM_011511908.2 XM_011511909.2 XM_011511910.1 XM_011511911.1 XM_011511912.1 XM_011511913.2 XM_011511914.2 XM_011511915.2 XM_011511916.2 XM_017005018.1 XM_017005019.1 XM_017005020.1 XM_017005021.1 XM_017005022.1 XM_017005023.1 XM_017005024.1 XM_017005025.1 XM_017005026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4583284 c.936A>T p.L312F Chr.2: 231259012-231259012 on Build GRCh38
5396465 c.941C>T p.P314L Chr.2: 231259017-231259017 on Build GRCh38
4573007 c.934T>A p.L312I Chr.2: 231259010-231259010 on Build GRCh38
6010702 c.1001C>T p.A334V Chr.2: 231259077-231259077 on Build GRCh38
3838834 c.1011G>A p.L337L Chr.2: 231259087-231259087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201981676 Chr.2: 231259012-231259012 on Build GRCh38
rs762055365 Chr.2: 231259063-231259063 on Build GRCh38
rs369454946 Chr.2: 231259081-231259081 on Build GRCh38
rs768182241 Chr.2: 231258949-231258949 on Build GRCh38
rs766609997 Chr.2: 231259092-231259092 on Build GRCh38
rs752253204 Chr.2: 231259006-231259006 on Build GRCh38
rs763607121 Chr.2: 231259139-231259139 on Build GRCh38
rs770566595 Chr.2: 231259027-231259027 on Build GRCh38
rs199986057 Chr.2: 231258965-231258965 on Build GRCh38
rs764909669 Chr.2: 231259099-231259099 on Build GRCh38
rs201247163 Chr.2: 231258961-231258961 on Build GRCh38
rs749422780 Chr.2: 231259051-231259051 on Build GRCh38
rs757960072 Chr.2: 231259128-231259128 on Build GRCh38
rs750816607 Chr.2: 231258957-231258957 on Build GRCh38
rs146386454 Chr.2: 231259045-231259045 on Build GRCh38
rs376952620 Chr.2: 231259067-231259067 on Build GRCh38
rs774544817 Chr.2: 231259056-231259056 on Build GRCh38
rs1021916557 Chr.2: 231258955-231258955 on Build GRCh38
rs959929161 Chr.2: 231259029-231259029 on Build GRCh38
rs897516876 Chr.2: 231259030-231259030 on Build GRCh38
rs35322543 Chr.2: 231259055-231259055 on Build GRCh38
rs758301318 Chr.2: 231259007-231259007 on Build GRCh38
rs952945086 Chr.2: 231259083-231259083 on Build GRCh38
rs141984986 Chr.2: 231259025-231259025 on Build GRCh38
rs753362239 Chr.2: 231258974-231258974 on Build GRCh38
rs567834789 Chr.2: 231258970-231258970 on Build GRCh38
rs761245119 Chr.2: 231259068-231259068 on Build GRCh38
rs754420501 Chr.2: 231258946-231258946 on Build GRCh38
rs751413482 Chr.2: 231259151-231259151 on Build GRCh38
rs774595749 Chr.2: 231258934-231258934 on Build GRCh38
rs777071719 Chr.2: 231259094-231259094 on Build GRCh38
rs752198367 Chr.2: 231259102-231259102 on Build GRCh38
rs761006081 Chr.2: 231258960-231258960 on Build GRCh38
rs759085044 Chr.2: 231259097-231259097 on Build GRCh38
rs908585186 Chr.2: 231259122-231259122 on Build GRCh38
rs759048563 Chr.2: 231259128-231259132 on Build GRCh38
rs778341994 Chr.2: 231258994-231258994 on Build GRCh38
rs754688653 Chr.2: 231258982-231258982 on Build GRCh38

View Full Product Details