Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56639029-56639300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5B
Entrez Gene ID
506
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide
Gene Aliases
ATPMB, ATPSB, HEL-S-271
UniGene
Hs.406510
Gene Chromosome Location
Chr.12: 56638175-56646068 on Build GRCh38
Transcripts
NM_001686.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7329602 c.1329G>T p.M443I Chr.12: 56639266-56639266 on Build GRCh38
4611596 c.1484_1485insT p.L495fs*4 Chr.12: 56639110-56639111 on Build GRCh38
292368 c.1376A>C p.K459T Chr.12: 56639219-56639219 on Build GRCh38
4954161 c.1395T>G p.S465S Chr.12: 56639200-56639200 on Build GRCh38
7317734 c.1466A>G p.K489R Chr.12: 56639129-56639129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746624237 Chr.12: 56639074-56639074 on Build GRCh38
rs113030570 Chr.12: 56639119-56639119 on Build GRCh38
rs767722996 Chr.12: 56639164-56639164 on Build GRCh38
rs770608613 Chr.12: 56639090-56639090 on Build GRCh38
rs145015204 Chr.12: 56639116-56639116 on Build GRCh38
rs888471798 Chr.12: 56639251-56639251 on Build GRCh38
rs149104052 Chr.12: 56639146-56639146 on Build GRCh38
rs758248124 Chr.12: 56639240-56639240 on Build GRCh38
rs772944609 Chr.12: 56639063-56639063 on Build GRCh38
rs970340533 Chr.12: 56639058-56639058 on Build GRCh38
rs750674096 Chr.12: 56639188-56639188 on Build GRCh38
rs776260481 Chr.12: 56639098-56639098 on Build GRCh38
rs762250167 Chr.12: 56639151-56639151 on Build GRCh38
rs374052119 Chr.12: 56639215-56639215 on Build GRCh38
rs926587967 Chr.12: 56639092-56639092 on Build GRCh38
rs760854189 Chr.12: 56639194-56639194 on Build GRCh38
rs778911180 Chr.12: 56639228-56639228 on Build GRCh38
rs199687768 Chr.12: 56639167-56639167 on Build GRCh38
rs1046598344 Chr.12: 56639075-56639075 on Build GRCh38
rs775394161 Chr.12: 56639254-56639254 on Build GRCh38
rs200610844 Chr.12: 56639211-56639211 on Build GRCh38
rs939384140 Chr.12: 56639109-56639109 on Build GRCh38
rs771709386 Chr.12: 56639060-56639060 on Build GRCh38
rs770694386 Chr.12: 56639245-56639245 on Build GRCh38
rs543239241 Chr.12: 56639173-56639173 on Build GRCh38
rs766490903 Chr.12: 56639210-56639210 on Build GRCh38
rs377482532 Chr.12: 56639223-56639223 on Build GRCh38
rs774311995 Chr.12: 56639129-56639129 on Build GRCh38
rs777684193 Chr.12: 56639242-56639242 on Build GRCh38
rs201918370 Chr.12: 56639170-56639170 on Build GRCh38
rs752530531 Chr.12: 56639236-56639236 on Build GRCh38
rs945550767 Chr.12: 56639069-56639069 on Build GRCh38
rs1005477882 Chr.12: 56639272-56639272 on Build GRCh38
rs746709831 Chr.12: 56639244-56639244 on Build GRCh38

View Full Product Details