Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27461678-27461947 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA2
Entrez Gene ID
1135
Gene Name
cholinergic receptor nicotinic alpha 2 subunit
Gene Aliases
-
UniGene
Hs.57718
Gene Chromosome Location
Chr.8: 27459761-27479296 on Build GRCh38
Transcripts
NM_000742.3 NM_001282455.1 XM_005273397.2 XM_006716282.1 XM_011544388.1 XM_011544389.1 XM_017013003.1 XM_017013004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7165700 c.1513C>T p.L505F Chr.8: 27461706-27461706 on Build GRCh38
3367305 c.1477T>C p.W493R Chr.8: 27461742-27461742 on Build GRCh38
4770743 c.1479delG p.W493fs*1 Chr.8: 27461740-27461740 on Build GRCh38
3834507 c.1466T>G p.V489G Chr.8: 27461753-27461753 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762003141 Chr.8: 27461738-27461738 on Build GRCh38
rs1044276848 Chr.8: 27461902-27461902 on Build GRCh38
rs796339838 Chr.8: 27461856-27461856 on Build GRCh38
rs569935505 Chr.8: 27461814-27461814 on Build GRCh38
rs996398612 Chr.8: 27461828-27461828 on Build GRCh38
rs747825857 Chr.8: 27461810-27461810 on Build GRCh38
rs759930402 Chr.8: 27461718-27461718 on Build GRCh38
rs1002825912 Chr.8: 27461923-27461923 on Build GRCh38
rs764727014 Chr.8: 27461782-27461782 on Build GRCh38
rs372045876 Chr.8: 27461776-27461776 on Build GRCh38
rs202078247 Chr.8: 27461799-27461799 on Build GRCh38
rs752601303 Chr.8: 27461774-27461774 on Build GRCh38
rs912849119 Chr.8: 27461841-27461841 on Build GRCh38
rs375095234 Chr.8: 27461737-27461737 on Build GRCh38
rs907152063 Chr.8: 27461905-27461905 on Build GRCh38
rs1027118146 Chr.8: 27461823-27461823 on Build GRCh38
rs529399073 Chr.8: 27461836-27461836 on Build GRCh38
rs549290936 Chr.8: 27461855-27461855 on Build GRCh38
rs776061785 Chr.8: 27461782-27461782 on Build GRCh38
rs767437489 Chr.8: 27461762-27461762 on Build GRCh38
rs1012797739 Chr.8: 27461703-27461703 on Build GRCh38
rs780391294 Chr.8: 27461798-27461798 on Build GRCh38
rs143223159 Chr.8: 27461719-27461719 on Build GRCh38
rs756820138 Chr.8: 27461781-27461781 on Build GRCh38
rs765805084 Chr.8: 27461749-27461751 on Build GRCh38
rs189738419 Chr.8: 27461901-27461901 on Build GRCh38
rs375469296 Chr.8: 27461757-27461757 on Build GRCh38
rs944338079 Chr.8: 27461842-27461842 on Build GRCh38
rs985704862 Chr.8: 27461837-27461837 on Build GRCh38
rs961479867 Chr.8: 27461783-27461783 on Build GRCh38
rs1030920374 Chr.8: 27461831-27461831 on Build GRCh38
rs758042994 Chr.8: 27461797-27461797 on Build GRCh38
rs746176483 Chr.8: 27461710-27461710 on Build GRCh38
rs755464771 Chr.8: 27461801-27461801 on Build GRCh38
rs777645463 Chr.8: 27461892-27461892 on Build GRCh38
rs533118782 Chr.8: 27461785-27461785 on Build GRCh38
rs772487934 Chr.8: 27461715-27461715 on Build GRCh38
rs370619269 Chr.8: 27461924-27461924 on Build GRCh38
rs147064296 Chr.8: 27461848-27461848 on Build GRCh38
rs735421 Chr.8: 27461775-27461775 on Build GRCh38
rs934183 Chr.8: 27461904-27461904 on Build GRCh38
rs534616396 Chr.8: 27461926-27461926 on Build GRCh38
rs975787699 Chr.8: 27461844-27461844 on Build GRCh38
rs760166123 Chr.8: 27461719-27461721 on Build GRCh38
rs773515773 Chr.8: 27461751-27461751 on Build GRCh38
rs374449470 Chr.8: 27461741-27461741 on Build GRCh38
rs924351510 Chr.8: 27461845-27461845 on Build GRCh38
rs747500158 Chr.8: 27461721-27461721 on Build GRCh38
rs202029606 Chr.8: 27461725-27461725 on Build GRCh38
rs954355898 Chr.8: 27461824-27461824 on Build GRCh38
rs752625951 Chr.8: 27461728-27461730 on Build GRCh38
rs186018525 Chr.8: 27461858-27461858 on Build GRCh38

View Full Product Details