Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30489481-30489754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-E
Entrez Gene ID
3133
Gene Name
major histocompatibility complex, class I, E
Gene Aliases
EA1.2, EA2.1, HLA-6.2, MHC, QA1
UniGene
Hs.650174
Gene Chromosome Location
Chr.6: 30489406-30494205 on Build GRCh38
Transcripts
NM_005516.5 XM_017010807.1 XM_017010808.1 XM_017010809.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1077338 c.75C>A p.S25S Chr.6: 30489736-30489736 on Build GRCh38
420917 c.30C>G p.L10L Chr.6: 30489561-30489561 on Build GRCh38
1634698 c.19C>T p.L7F Chr.6: 30489550-30489550 on Build GRCh38
5495521 c.15C>T p.T5T Chr.6: 30489546-30489546 on Build GRCh38
6221340 c.18C>T p.L6L Chr.6: 30489549-30489549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779331330 Chr.6: 30489509-30489509 on Build GRCh38
rs369586994 Chr.6: 30489577-30489577 on Build GRCh38
rs375999164 Chr.6: 30489689-30489689 on Build GRCh38
rs76971248 Chr.6: 30489506-30489506 on Build GRCh38
rs758950539 Chr.6: 30489514-30489514 on Build GRCh38
rs759003552 Chr.6: 30489596-30489596 on Build GRCh38
rs937920143 Chr.6: 30489519-30489519 on Build GRCh38
rs1039160819 Chr.6: 30489624-30489624 on Build GRCh38
rs778020430 Chr.6: 30489517-30489517 on Build GRCh38
rs368803466 Chr.6: 30489531-30489531 on Build GRCh38
rs112472308 Chr.6: 30489724-30489724 on Build GRCh38
rs376826280 Chr.6: 30489550-30489550 on Build GRCh38
rs890202908 Chr.6: 30489587-30489587 on Build GRCh38
rs369524967 Chr.6: 30489547-30489547 on Build GRCh38
rs200086098 Chr.6: 30489639-30489639 on Build GRCh38
rs746962356 Chr.6: 30489668-30489668 on Build GRCh38
rs767067348 Chr.6: 30489501-30489501 on Build GRCh38
rs759133831 Chr.6: 30489681-30489681 on Build GRCh38
rs763348367 Chr.6: 30489684-30489684 on Build GRCh38
rs776000632 Chr.6: 30489683-30489683 on Build GRCh38
rs753501391 Chr.6: 30489576-30489576 on Build GRCh38
rs761765470 Chr.6: 30489541-30489541 on Build GRCh38
rs937558103 Chr.6: 30489539-30489539 on Build GRCh38
rs372172747 Chr.6: 30489542-30489542 on Build GRCh38
rs760122302 Chr.6: 30489549-30489549 on Build GRCh38
rs368589350 Chr.6: 30489502-30489502 on Build GRCh38
rs772012998 Chr.6: 30489653-30489653 on Build GRCh38
rs776182677 Chr.6: 30489672-30489672 on Build GRCh38
rs113161579 Chr.6: 30489511-30489511 on Build GRCh38
rs764941140 Chr.6: 30489682-30489682 on Build GRCh38
rs764744109 Chr.6: 30489512-30489512 on Build GRCh38
rs749920862 Chr.6: 30489656-30489656 on Build GRCh38
rs770388769 Chr.6: 30489669-30489669 on Build GRCh38
rs55785318 Chr.6: 30489560-30489560 on Build GRCh38
rs753061470 Chr.6: 30489583-30489583 on Build GRCh38
rs746086512 Chr.6: 30489532-30489532 on Build GRCh38
rs774114493 Chr.6: 30489538-30489538 on Build GRCh38
rs751700263 Chr.6: 30489620-30489620 on Build GRCh38
rs765570299 Chr.6: 30489582-30489582 on Build GRCh38
rs778715813 Chr.6: 30489649-30489649 on Build GRCh38
rs757747376 Chr.6: 30489687-30489687 on Build GRCh38
rs761434219 Chr.6: 30489518-30489519 on Build GRCh38
rs534874909 Chr.6: 30489609-30489609 on Build GRCh38
rs764538201 Chr.6: 30489685-30489685 on Build GRCh38
rs772715533 Chr.6: 30489545-30489545 on Build GRCh38
rs768398044 Chr.6: 30489535-30489535 on Build GRCh38
rs752174313 Chr.6: 30489686-30489686 on Build GRCh38
rs747043330 Chr.6: 30489524-30489524 on Build GRCh38
rs200004109 Chr.6: 30489528-30489528 on Build GRCh38
rs757337088 Chr.6: 30489626-30489626 on Build GRCh38
rs754243872 Chr.6: 30489503-30489503 on Build GRCh38
rs17875368 Chr.6: 30489652-30489652 on Build GRCh38
rs781324002 Chr.6: 30489631-30489631 on Build GRCh38
rs750480675 Chr.6: 30489700-30489700 on Build GRCh38

View Full Product Details