Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 94461999-94462234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCT
Entrez Gene ID
1638
Gene Name
dopachrome tautomerase
Gene Aliases
TRP-2, TYRP2
UniGene
Hs.301865
Gene Chromosome Location
Chr.13: 94437304-94550265 on Build GRCh38
Transcripts
NM_001129889.2 NM_001322182.1 NM_001322183.1 NM_001322184.1 NM_001322185.1 NM_001322186.1 NM_001922.4 XM_011521049.1 XM_017020401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4641833 c.876C>A p.Y292* Chr.13: 94462177-94462177 on Build GRCh38
3793413 c.864-2A>T p.? Chr.13: 94462191-94462191 on Build GRCh38
6537996 c.951G>C p.M317I Chr.13: 94462102-94462102 on Build GRCh38
1629187 c.1043+5G>C p.? Chr.13: 94462005-94462005 on Build GRCh38
1629188 c.1043+5G>C p.? Chr.13: 94462005-94462005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776517934 Chr.13: 94462084-94462084 on Build GRCh38
rs757604658 Chr.13: 94462051-94462051 on Build GRCh38
rs760281161 Chr.13: 94462127-94462127 on Build GRCh38
rs74325354 Chr.13: 94462144-94462144 on Build GRCh38
rs781489627 Chr.13: 94462069-94462069 on Build GRCh38
rs758950858 Chr.13: 94462048-94462048 on Build GRCh38
rs140917943 Chr.13: 94462076-94462076 on Build GRCh38
rs753213570 Chr.13: 94462046-94462046 on Build GRCh38
rs750637872 Chr.13: 94462186-94462186 on Build GRCh38
rs375498582 Chr.13: 94462103-94462103 on Build GRCh38
rs1011900553 Chr.13: 94462066-94462066 on Build GRCh38
rs142117845 Chr.13: 94462068-94462068 on Build GRCh38
rs763407563 Chr.13: 94462145-94462145 on Build GRCh38
rs756039485 Chr.13: 94462108-94462108 on Build GRCh38
rs751862593 Chr.13: 94462171-94462171 on Build GRCh38
rs764587115 Chr.13: 94462169-94462169 on Build GRCh38
rs772918072 Chr.13: 94462120-94462120 on Build GRCh38
rs1029675720 Chr.13: 94462185-94462185 on Build GRCh38
rs201692579 Chr.13: 94462177-94462177 on Build GRCh38
rs768747820 Chr.13: 94462081-94462081 on Build GRCh38
rs747973265 Chr.13: 94462101-94462101 on Build GRCh38
rs778060068 Chr.13: 94462049-94462049 on Build GRCh38
rs751950305 Chr.13: 94462067-94462067 on Build GRCh38
rs988866489 Chr.13: 94462166-94462166 on Build GRCh38
rs956502652 Chr.13: 94462157-94462157 on Build GRCh38
rs756314626 Chr.13: 94462191-94462191 on Build GRCh38
rs746133761 Chr.13: 94462071-94462071 on Build GRCh38
rs1023580929 Chr.13: 94462070-94462070 on Build GRCh38
rs767812669 Chr.13: 94462178-94462178 on Build GRCh38
rs774224411 Chr.13: 94462095-94462095 on Build GRCh38
rs749724235 Chr.13: 94462019-94462019 on Build GRCh38
rs367891461 Chr.13: 94462136-94462136 on Build GRCh38
rs148575940 Chr.13: 94462077-94462077 on Build GRCh38
rs935027499 Chr.13: 94462135-94462135 on Build GRCh38

View Full Product Details