Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 22225647-22225916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF2K
Entrez Gene ID
29904
Gene Name
eukaryotic elongation factor 2 kinase
Gene Aliases
HSU93850, eEF-2K
UniGene
Hs.498892
Gene Chromosome Location
Chr.16: 22206264-22288745 on Build GRCh38
Transcripts
NM_013302.3 XM_017023193.1 XM_017023194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3507624 c.144G>A p.S48S Chr.16: 22225873-22225873 on Build GRCh38
302115 c.122C>A p.P41H Chr.16: 22225851-22225851 on Build GRCh38
3706937 c.158A>G p.N53S Chr.16: 22225887-22225887 on Build GRCh38
4808253 c.26G>A p.R9H Chr.16: 22225755-22225755 on Build GRCh38
5473712 c.94G>A p.D32N Chr.16: 22225823-22225823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759185801 Chr.16: 22225816-22225816 on Build GRCh38
rs752397121 Chr.16: 22225755-22225755 on Build GRCh38
rs1011048154 Chr.16: 22225714-22225714 on Build GRCh38
rs762230195 Chr.16: 22225696-22225696 on Build GRCh38
rs954169807 Chr.16: 22225824-22225824 on Build GRCh38
rs765834492 Chr.16: 22225731-22225731 on Build GRCh38
rs749205317 Chr.16: 22225766-22225766 on Build GRCh38
rs763827414 Chr.16: 22225835-22225835 on Build GRCh38
rs866141083 Chr.16: 22225787-22225787 on Build GRCh38
rs9935059 Chr.16: 22225797-22225797 on Build GRCh38
rs751118481 Chr.16: 22225738-22225738 on Build GRCh38
rs982180183 Chr.16: 22225836-22225836 on Build GRCh38
rs748311349 Chr.16: 22225786-22225786 on Build GRCh38
rs747148020 Chr.16: 22225776-22225776 on Build GRCh38
rs770090956 Chr.16: 22225865-22225865 on Build GRCh38
rs377003950 Chr.16: 22225817-22225817 on Build GRCh38
rs965379418 Chr.16: 22225799-22225799 on Build GRCh38
rs776843094 Chr.16: 22225784-22225784 on Build GRCh38
rs773581150 Chr.16: 22225795-22225795 on Build GRCh38
rs145675806 Chr.16: 22225718-22225718 on Build GRCh38
rs781199038 Chr.16: 22225857-22225857 on Build GRCh38
rs756982467 Chr.16: 22225841-22225841 on Build GRCh38
rs201901191 Chr.16: 22225869-22225869 on Build GRCh38
rs773605025 Chr.16: 22225724-22225724 on Build GRCh38
rs142308079 Chr.16: 22225791-22225791 on Build GRCh38
rs778974609 Chr.16: 22225843-22225843 on Build GRCh38
rs554551621 Chr.16: 22225676-22225676 on Build GRCh38
rs753709983 Chr.16: 22225839-22225839 on Build GRCh38
rs768907973 Chr.16: 22225692-22225692 on Build GRCh38
rs191219230 Chr.16: 22225748-22225748 on Build GRCh38
rs750292606 Chr.16: 22225844-22225844 on Build GRCh38
rs755064229 Chr.16: 22225851-22225851 on Build GRCh38
rs148266060 Chr.16: 22225822-22225822 on Build GRCh38
rs777567285 Chr.16: 22225765-22225765 on Build GRCh38
rs774988381 Chr.16: 22225815-22225815 on Build GRCh38
rs949510226 Chr.16: 22225834-22225834 on Build GRCh38
rs766955813 Chr.16: 22225802-22225802 on Build GRCh38
rs748395567 Chr.16: 22225858-22225858 on Build GRCh38
rs748539609 Chr.16: 22225840-22225842 on Build GRCh38
rs760356178 Chr.16: 22225825-22225825 on Build GRCh38
rs764501820 Chr.16: 22225737-22225737 on Build GRCh38
rs146984360 Chr.16: 22225754-22225754 on Build GRCh38
rs866336266 Chr.16: 22225863-22225863 on Build GRCh38
rs778685874 Chr.16: 22225775-22225775 on Build GRCh38
rs529625938 Chr.16: 22225873-22225873 on Build GRCh38
rs757180437 Chr.16: 22225774-22225774 on Build GRCh38
rs763465488 Chr.16: 22225730-22225730 on Build GRCh38
rs147612457 Chr.16: 22225783-22225783 on Build GRCh38
rs574420667 Chr.16: 22225694-22225694 on Build GRCh38
rs201562293 Chr.16: 22225887-22225887 on Build GRCh38

View Full Product Details