Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65115667-65115924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TM7SF2
Entrez Gene ID
7108
Gene Name
transmembrane 7 superfamily member 2
Gene Aliases
ANG1, DHCR14A, NET47
UniGene
Hs.31130
Gene Chromosome Location
Chr.11: 65111854-65116235 on Build GRCh38
Transcripts
NM_001277233.1 NM_003273.3 NR_102367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
429585 c.1111C>T p.L371L Chr.11: 65115907-65115907 on Build GRCh38
3788024 c.1117T>C p.Y373H Chr.11: 65115913-65115913 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769613136 Chr.11: 65115774-65115774 on Build GRCh38
rs369504487 Chr.11: 65115847-65115847 on Build GRCh38
rs1031508906 Chr.11: 65115702-65115702 on Build GRCh38
rs376755166 Chr.11: 65115846-65115846 on Build GRCh38
rs749953666 Chr.11: 65115892-65115892 on Build GRCh38
rs750967884 Chr.11: 65115737-65115737 on Build GRCh38
rs111853188 Chr.11: 65115891-65115891 on Build GRCh38
rs760854071 Chr.11: 65115811-65115811 on Build GRCh38
rs868480128 Chr.11: 65115778-65115778 on Build GRCh38
rs571392774 Chr.11: 65115699-65115700 on Build GRCh38
rs772212746 Chr.11: 65115786-65115786 on Build GRCh38
rs916323416 Chr.11: 65115787-65115787 on Build GRCh38
rs766297848 Chr.11: 65115758-65115758 on Build GRCh38
rs993140511 Chr.11: 65115725-65115725 on Build GRCh38
rs960008423 Chr.11: 65115721-65115721 on Build GRCh38
rs929390058 Chr.11: 65115792-65115792 on Build GRCh38
rs537944472 Chr.11: 65115690-65115690 on Build GRCh38
rs780636053 Chr.11: 65115811-65115812 on Build GRCh38
rs557757637 Chr.11: 65115694-65115694 on Build GRCh38
rs774923650 Chr.11: 65115781-65115781 on Build GRCh38
rs138553302 Chr.11: 65115688-65115688 on Build GRCh38
rs534168939 Chr.11: 65115860-65115860 on Build GRCh38
rs762742172 Chr.11: 65115819-65115819 on Build GRCh38
rs749593352 Chr.11: 65115850-65115850 on Build GRCh38
rs777754429 Chr.11: 65115731-65115731 on Build GRCh38
rs749719529 Chr.11: 65115871-65115872 on Build GRCh38
rs749732978 Chr.11: 65115777-65115777 on Build GRCh38
rs776853364 Chr.11: 65115869-65115869 on Build GRCh38
rs112104203 Chr.11: 65115890-65115890 on Build GRCh38
rs761738080 Chr.11: 65115782-65115782 on Build GRCh38
rs779236284 Chr.11: 65115858-65115858 on Build GRCh38
rs780661792 Chr.11: 65115768-65115768 on Build GRCh38
rs755404638 Chr.11: 65115854-65115854 on Build GRCh38
rs774056804 Chr.11: 65115887-65115887 on Build GRCh38
rs763983694 Chr.11: 65115877-65115877 on Build GRCh38
rs756805867 Chr.11: 65115750-65115750 on Build GRCh38
rs762688993 Chr.11: 65115875-65115875 on Build GRCh38
rs544507657 Chr.11: 65115903-65115903 on Build GRCh38
rs577942022 Chr.11: 65115845-65115845 on Build GRCh38
rs554173160 Chr.11: 65115865-65115865 on Build GRCh38
rs373673390 Chr.11: 65115849-65115849 on Build GRCh38
rs747149960 Chr.11: 65115866-65115866 on Build GRCh38
rs752527409 Chr.11: 65115727-65115727 on Build GRCh38
rs746496230 Chr.11: 65115757-65115757 on Build GRCh38
rs766636203 Chr.11: 65115813-65115813 on Build GRCh38
rs1012043850 Chr.11: 65115893-65115893 on Build GRCh38
rs770255839 Chr.11: 65115772-65115777 on Build GRCh38
rs773263887 Chr.11: 65115810-65115810 on Build GRCh38
rs780547087 Chr.11: 65115776-65115776 on Build GRCh38
rs758309629 Chr.11: 65115728-65115728 on Build GRCh38
rs375239318 Chr.11: 65115895-65115895 on Build GRCh38
rs771075042 Chr.11: 65115867-65115867 on Build GRCh38
rs752484564 Chr.11: 65115814-65115814 on Build GRCh38
rs769277354 Chr.11: 65115779-65115779 on Build GRCh38
rs772326114 Chr.11: 65115864-65115864 on Build GRCh38

View Full Product Details