Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78187236-78187461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFMID
Entrez Gene ID
125061
Gene Name
arylformamidase
Gene Aliases
FKF, KF, KFA
UniGene
Hs.558614
Gene Chromosome Location
Chr.17: 78185102-78207702 on Build GRCh38
Transcripts
NM_001010982.4 NM_001145526.2 NR_027083.1 XM_011524326.2 XM_011524327.2 XM_011524328.2 XM_011524329.2 XM_011524330.2 XM_011524331.2 XM_011524332.2 XM_011524333.2 XM_011524334.2 XM_011524335.2 XM_011524336.2 XM_011524337.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4910804 c.1A>G p.M1V Chr.17: 78187371-78187371 on Build GRCh38
3522786 c.7G>A p.D3N Chr.17: 78187377-78187377 on Build GRCh38
2745681 c.45_47delGAA p.K17delK Chr.17: 78187415-78187417 on Build GRCh38
4580185 c.14C>G p.S5C Chr.17: 78187384-78187384 on Build GRCh38
4919584 c.51G>A p.K17K Chr.17: 78187421-78187421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368272932 Chr.17: 78187411-78187411 on Build GRCh38
rs1013553604 Chr.17: 78187352-78187352 on Build GRCh38
rs964630527 Chr.17: 78187433-78187433 on Build GRCh38
rs770995525 Chr.17: 78187412-78187412 on Build GRCh38
rs878875265 Chr.17: 78187314-78187314 on Build GRCh38
rs764106366 Chr.17: 78187350-78187350 on Build GRCh38
rs878887356 Chr.17: 78187317-78187317 on Build GRCh38
rs767731459 Chr.17: 78187377-78187377 on Build GRCh38
rs1020537109 Chr.17: 78187437-78187437 on Build GRCh38
rs749299277 Chr.17: 78187366-78187366 on Build GRCh38
rs371795903 Chr.17: 78187367-78187367 on Build GRCh38
rs910496957 Chr.17: 78187286-78187286 on Build GRCh38
rs777252843 Chr.17: 78187322-78187322 on Build GRCh38
rs375000062 Chr.17: 78187341-78187341 on Build GRCh38
rs971569135 Chr.17: 78187263-78187263 on Build GRCh38
rs774544979 Chr.17: 78187370-78187370 on Build GRCh38
rs1031790335 Chr.17: 78187382-78187382 on Build GRCh38
rs752967953 Chr.17: 78187406-78187406 on Build GRCh38
rs748546152 Chr.17: 78187357-78187357 on Build GRCh38
rs118063987 Chr.17: 78187267-78187267 on Build GRCh38
rs753237566 Chr.17: 78187353-78187353 on Build GRCh38
rs370147226 Chr.17: 78187330-78187330 on Build GRCh38
rs778897924 Chr.17: 78187420-78187420 on Build GRCh38
rs767359518 Chr.17: 78187348-78187348 on Build GRCh38
rs761204375 Chr.17: 78187394-78187394 on Build GRCh38
rs773803862 Chr.17: 78187364-78187364 on Build GRCh38
rs763118964 Chr.17: 78187332-78187332 on Build GRCh38
rs775209283 Chr.17: 78187384-78187384 on Build GRCh38
rs368006213 Chr.17: 78187347-78187347 on Build GRCh38
rs756545065 Chr.17: 78187409-78187409 on Build GRCh38
rs937061427 Chr.17: 78187302-78187303 on Build GRCh38
rs748719252 Chr.17: 78187328-78187328 on Build GRCh38
rs756822084 Chr.17: 78187354-78187354 on Build GRCh38
rs769908844 Chr.17: 78187329-78187329 on Build GRCh38
rs998964915 Chr.17: 78187379-78187379 on Build GRCh38
rs775051254 Chr.17: 78187385-78187401 on Build GRCh38
rs368094951 Chr.17: 78187296-78187296 on Build GRCh38
rs745562416 Chr.17: 78187356-78187356 on Build GRCh38
rs943265598 Chr.17: 78187300-78187300 on Build GRCh38
rs756671596 Chr.17: 78187395-78187395 on Build GRCh38
rs763896884 Chr.17: 78187388-78187388 on Build GRCh38
rs778674864 Chr.17: 78187355-78187355 on Build GRCh38
rs753852393 Chr.17: 78187390-78187390 on Build GRCh38
rs368543891 Chr.17: 78187320-78187320 on Build GRCh38
rs771869590 Chr.17: 78187338-78187339 on Build GRCh38
rs373566532 Chr.17: 78187262-78187263 on Build GRCh38
rs752627458 Chr.17: 78187349-78187349 on Build GRCh38
rs759823605 Chr.17: 78187372-78187372 on Build GRCh38
rs760541616 Chr.17: 78187387-78187387 on Build GRCh38
rs929073009 Chr.17: 78187321-78187321 on Build GRCh38
rs766473569 Chr.17: 78187336-78187336 on Build GRCh38
rs764895979 Chr.17: 78187399-78187399 on Build GRCh38
rs746638950 Chr.17: 78187415-78187417 on Build GRCh38
rs745962259 Chr.17: 78187421-78187421 on Build GRCh38
rs371981367 Chr.17: 78187359-78187359 on Build GRCh38
rs1040072098 Chr.17: 78187302-78187302 on Build GRCh38
rs770155744 Chr.17: 78187362-78187362 on Build GRCh38
rs927016861 Chr.17: 78187297-78187297 on Build GRCh38
rs376426687 Chr.17: 78187400-78187400 on Build GRCh38
rs779878281 Chr.17: 78187402-78187402 on Build GRCh38

View Full Product Details