Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 33013012-33013239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK2
Entrez Gene ID
204
Gene Name
adenylate kinase 2
Gene Aliases
ADK2
UniGene
Hs.470907
Gene Chromosome Location
Chr.1: 33007940-33036911 on Build GRCh38
Transcripts
NM_001199199.1 NM_001319139.1 NM_001319140.1 NM_001319141.1 NM_001319142.1 NM_001319143.1 NM_001625.3 NM_013411.4 NR_134976.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3943878 c.700G>C p.D234H Chr.1: 33013201-33013201 on Build GRCh38
6054480 c.668T>C p.I223T Chr.1: 33013233-33013233 on Build GRCh38
4399250 c.684C>T p.S228S Chr.1: 33013217-33013217 on Build GRCh38
4399251 c.684C>T p.S228S Chr.1: 33013217-33013217 on Build GRCh38
3943879 c.694+6G>C p.? Chr.1: 33013201-33013201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752483572 Chr.1: 33013052-33013052 on Build GRCh38
rs767186739 Chr.1: 33013049-33013049 on Build GRCh38
rs770143873 Chr.1: 33013117-33013117 on Build GRCh38
rs771662385 Chr.1: 33013146-33013146 on Build GRCh38
rs183996331 Chr.1: 33013035-33013035 on Build GRCh38
rs748489922 Chr.1: 33013098-33013098 on Build GRCh38
rs191407523 Chr.1: 33013167-33013167 on Build GRCh38
rs760938958 Chr.1: 33013037-33013037 on Build GRCh38
rs937755989 Chr.1: 33013124-33013124 on Build GRCh38
rs145687281 Chr.1: 33013138-33013138 on Build GRCh38
rs752317962 Chr.1: 33013082-33013082 on Build GRCh38
rs372620503 Chr.1: 33013148-33013148 on Build GRCh38
rs767017313 Chr.1: 33013040-33013040 on Build GRCh38
rs760411755 Chr.1: 33013158-33013158 on Build GRCh38
rs370310306 Chr.1: 33013136-33013136 on Build GRCh38
rs752903099 Chr.1: 33013191-33013191 on Build GRCh38
rs751725148 Chr.1: 33013182-33013182 on Build GRCh38
rs995686582 Chr.1: 33013046-33013046 on Build GRCh38
rs768207363 Chr.1: 33013179-33013179 on Build GRCh38
rs1034726716 Chr.1: 33013089-33013089 on Build GRCh38
rs756443428 Chr.1: 33013193-33013193 on Build GRCh38
rs749644551 Chr.1: 33013208-33013208 on Build GRCh38
rs370479789 Chr.1: 33013120-33013120 on Build GRCh38
rs527257332 Chr.1: 33013060-33013060 on Build GRCh38
rs375860853 Chr.1: 33013181-33013181 on Build GRCh38
rs763970731 Chr.1: 33013059-33013059 on Build GRCh38
rs188290204 Chr.1: 33013083-33013083 on Build GRCh38
rs771479963 Chr.1: 33013218-33013218 on Build GRCh38
rs761900027 Chr.1: 33013180-33013180 on Build GRCh38
rs755908960 Chr.1: 33013053-33013053 on Build GRCh38
rs780348453 Chr.1: 33013071-33013071 on Build GRCh38
rs960735924 Chr.1: 33013092-33013092 on Build GRCh38
rs267606646 Chr.1: 33013204-33013204 on Build GRCh38
rs139639834 Chr.1: 33013187-33013187 on Build GRCh38
rs762469339 Chr.1: 33013036-33013036 on Build GRCh38
rs553945682 Chr.1: 33013186-33013186 on Build GRCh38
rs751675835 Chr.1: 33013077-33013077 on Build GRCh38
rs759255603 Chr.1: 33013048-33013048 on Build GRCh38
rs758554149 Chr.1: 33013069-33013069 on Build GRCh38
rs770073662 Chr.1: 33013195-33013195 on Build GRCh38
rs753769060 Chr.1: 33013065-33013065 on Build GRCh38
rs778269600 Chr.1: 33013199-33013199 on Build GRCh38
rs868380353 Chr.1: 33013202-33013202 on Build GRCh38
rs778158669 Chr.1: 33013119-33013119 on Build GRCh38

View Full Product Details