Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 94465584-94465851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCT
Entrez Gene ID
1638
Gene Name
dopachrome tautomerase
Gene Aliases
TRP-2, TYRP2
UniGene
Hs.301865
Gene Chromosome Location
Chr.13: 94437304-94550265 on Build GRCh38
Transcripts
NM_001129889.2 NM_001322182.1 NM_001322183.1 NM_001322184.1 NM_001322185.1 NM_001322186.1 NM_001922.4 XM_011521049.1 XM_017020401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3360157 c.823C>T p.R275W Chr.13: 94465673-94465673 on Build GRCh38
4556904 c.706G>A p.G236S Chr.13: 94465790-94465790 on Build GRCh38
4676603 c.790_791insG p.A264fs*11 Chr.13: 94465705-94465706 on Build GRCh38
6493813 c.753delG p.N252fs*21 Chr.13: 94465743-94465743 on Build GRCh38
4560960 c.863G>A p.S288N Chr.13: 94465633-94465633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746294793 Chr.13: 94465616-94465622 on Build GRCh38
rs762340057 Chr.13: 94465744-94465744 on Build GRCh38
rs745473948 Chr.13: 94465817-94465817 on Build GRCh38
rs754908817 Chr.13: 94465611-94465611 on Build GRCh38
rs200220737 Chr.13: 94465687-94465687 on Build GRCh38
rs201937888 Chr.13: 94465646-94465646 on Build GRCh38
rs1008077150 Chr.13: 94465701-94465701 on Build GRCh38
rs775382645 Chr.13: 94465682-94465683 on Build GRCh38
rs764784624 Chr.13: 94465649-94465649 on Build GRCh38
rs746620620 Chr.13: 94465633-94465633 on Build GRCh38
rs142171852 Chr.13: 94465691-94465691 on Build GRCh38
rs139109752 Chr.13: 94465655-94465655 on Build GRCh38
rs548383203 Chr.13: 94465792-94465792 on Build GRCh38
rs368402070 Chr.13: 94465616-94465616 on Build GRCh38
rs148902424 Chr.13: 94465761-94465761 on Build GRCh38
rs147581121 Chr.13: 94465686-94465686 on Build GRCh38
rs769271753 Chr.13: 94465740-94465740 on Build GRCh38
rs754077356 Chr.13: 94465782-94465782 on Build GRCh38
rs943910567 Chr.13: 94465749-94465749 on Build GRCh38
rs967496122 Chr.13: 94465762-94465762 on Build GRCh38
rs777581598 Chr.13: 94465706-94465706 on Build GRCh38
rs760932056 Chr.13: 94465769-94465769 on Build GRCh38
rs1015165708 Chr.13: 94465621-94465621 on Build GRCh38
rs769466143 Chr.13: 94465822-94465822 on Build GRCh38
rs770558686 Chr.13: 94465723-94465723 on Build GRCh38
rs759890417 Chr.13: 94465789-94465789 on Build GRCh38
rs766914313 Chr.13: 94465776-94465776 on Build GRCh38
rs758286477 Chr.13: 94465698-94465698 on Build GRCh38
rs34139115 Chr.13: 94465672-94465672 on Build GRCh38
rs977096912 Chr.13: 94465763-94465763 on Build GRCh38
rs145666217 Chr.13: 94465741-94465741 on Build GRCh38
rs776050229 Chr.13: 94465647-94465647 on Build GRCh38
rs112538123 Chr.13: 94465797-94465797 on Build GRCh38
rs765362254 Chr.13: 94465791-94465791 on Build GRCh38
rs373529464 Chr.13: 94465807-94465807 on Build GRCh38
rs772925819 Chr.13: 94465628-94465628 on Build GRCh38
rs759059356 Chr.13: 94465648-94465648 on Build GRCh38
rs771865026 Chr.13: 94465622-94465622 on Build GRCh38
rs774835333 Chr.13: 94465653-94465653 on Build GRCh38
rs199868596 Chr.13: 94465765-94465765 on Build GRCh38
rs558315058 Chr.13: 94465643-94465643 on Build GRCh38
rs200698216 Chr.13: 94465656-94465656 on Build GRCh38
rs748167345 Chr.13: 94465618-94465618 on Build GRCh38
rs369436840 Chr.13: 94465799-94465799 on Build GRCh38
rs766526063 Chr.13: 94465673-94465673 on Build GRCh38
rs772222841 Chr.13: 94465644-94465644 on Build GRCh38
rs1040889024 Chr.13: 94465768-94465768 on Build GRCh38
rs981625388 Chr.13: 94465663-94465663 on Build GRCh38
rs780808126 Chr.13: 94465725-94465725 on Build GRCh38
rs144365832 Chr.13: 94465739-94465739 on Build GRCh38
rs201510532 Chr.13: 94465692-94465692 on Build GRCh38
rs377082349 Chr.13: 94465758-94465758 on Build GRCh38
rs750785411 Chr.13: 94465662-94465662 on Build GRCh38
rs143672390 Chr.13: 94465798-94465798 on Build GRCh38
rs746782953 Chr.13: 94465713-94465713 on Build GRCh38
rs150712681 Chr.13: 94465640-94465640 on Build GRCh38
rs758291069 Chr.13: 94465793-94465793 on Build GRCh38

View Full Product Details