Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56639239-56639464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5B
Entrez Gene ID
506
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide
Gene Aliases
ATPMB, ATPSB, HEL-S-271
UniGene
Hs.406510
Gene Chromosome Location
Chr.12: 56638175-56646068 on Build GRCh38
Transcripts
NM_001686.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7329602 c.1329G>T p.M443I Chr.12: 56639266-56639266 on Build GRCh38
4825039 c.1288-5C>T p.? Chr.12: 56639312-56639312 on Build GRCh38
127393 c.1320C>T p.I440I Chr.12: 56639275-56639275 on Build GRCh38
941790 c.1349A>G p.D450G Chr.12: 56639246-56639246 on Build GRCh38
4525646 c.1344G>A p.E448E Chr.12: 56639251-56639251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780918856 Chr.12: 56639296-56639296 on Build GRCh38
rs754729472 Chr.12: 56639319-56639321 on Build GRCh38
rs745377010 Chr.12: 56639300-56639300 on Build GRCh38
rs1007313369 Chr.12: 56639395-56639395 on Build GRCh38
rs1030158523 Chr.12: 56639381-56639381 on Build GRCh38
rs369157556 Chr.12: 56639346-56639346 on Build GRCh38
rs774942479 Chr.12: 56639312-56639312 on Build GRCh38
rs773538892 Chr.12: 56639437-56639437 on Build GRCh38
rs773540539 Chr.12: 56639354-56639354 on Build GRCh38
rs536057742 Chr.12: 56639368-56639368 on Build GRCh38
rs371002521 Chr.12: 56639311-56639311 on Build GRCh38
rs1043838413 Chr.12: 56639330-56639330 on Build GRCh38
rs557558731 Chr.12: 56639425-56639425 on Build GRCh38
rs762187472 Chr.12: 56639316-56639316 on Build GRCh38
rs772526677 Chr.12: 56639340-56639340 on Build GRCh38
rs890254235 Chr.12: 56639393-56639393 on Build GRCh38
rs772491478 Chr.12: 56639410-56639410 on Build GRCh38
rs748228519 Chr.12: 56639366-56639366 on Build GRCh38
rs769127457 Chr.12: 56639307-56639307 on Build GRCh38
rs760941781 Chr.12: 56639356-56639356 on Build GRCh38
rs34004174 Chr.12: 56639355-56639356 on Build GRCh38
rs904021336 Chr.12: 56639389-56639389 on Build GRCh38
rs1005477882 Chr.12: 56639272-56639272 on Build GRCh38

View Full Product Details