Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69168177-69168402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNB1
Entrez Gene ID
891
Gene Name
cyclin B1
Gene Aliases
CCNB
UniGene
Hs.23960
Gene Chromosome Location
Chr.5: 69167010-69178245 on Build GRCh38
Transcripts
NM_031966.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1069677 c.245C>A p.P82H Chr.5: 69168225-69168225 on Build GRCh38
5014497 c.319C>T p.P107S Chr.5: 69168299-69168299 on Build GRCh38
1069678 c.344_345delTT p.S116fs*2 Chr.5: 69168324-69168325 on Build GRCh38
3072053 c.339A>C p.E113D Chr.5: 69168319-69168319 on Build GRCh38
1620489 c.210T>G p.A70A Chr.5: 69168190-69168190 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs527515881 Chr.5: 69168278-69168289 on Build GRCh38
rs755827930 Chr.5: 69168258-69168258 on Build GRCh38
rs761646129 Chr.5: 69168328-69168328 on Build GRCh38
rs920947659 Chr.5: 69168208-69168208 on Build GRCh38
rs780419001 Chr.5: 69168256-69168267 on Build GRCh38
rs779896517 Chr.5: 69168272-69168272 on Build GRCh38
rs776785541 Chr.5: 69168321-69168321 on Build GRCh38
rs760811100 Chr.5: 69168299-69168299 on Build GRCh38
rs757225235 Chr.5: 69168229-69168229 on Build GRCh38
rs780512901 Chr.5: 69168235-69168235 on Build GRCh38
rs751415136 Chr.5: 69168225-69168225 on Build GRCh38
rs867133028 Chr.5: 69168270-69168270 on Build GRCh38
rs748534626 Chr.5: 69168292-69168292 on Build GRCh38
rs755737995 Chr.5: 69168353-69168353 on Build GRCh38
rs763731070 Chr.5: 69168327-69168327 on Build GRCh38
rs768849718 Chr.5: 69168288-69168288 on Build GRCh38
rs771109821 Chr.5: 69168302-69168302 on Build GRCh38
rs183314174 Chr.5: 69168341-69168341 on Build GRCh38
rs758742437 Chr.5: 69168250-69168267 on Build GRCh38
rs954971463 Chr.5: 69168242-69168242 on Build GRCh38
rs892418645 Chr.5: 69168308-69168308 on Build GRCh38
rs936308097 Chr.5: 69168215-69168215 on Build GRCh38
rs779376687 Chr.5: 69168242-69168243 on Build GRCh38
rs147065029 Chr.5: 69168289-69168289 on Build GRCh38
rs376960487 Chr.5: 69168335-69168335 on Build GRCh38
rs906337231 Chr.5: 69168368-69168368 on Build GRCh38
rs750014957 Chr.5: 69168240-69168240 on Build GRCh38
rs374129207 Chr.5: 69168203-69168203 on Build GRCh38
rs749645134 Chr.5: 69168286-69168286 on Build GRCh38
rs773265958 Chr.5: 69168298-69168298 on Build GRCh38
rs144048886 Chr.5: 69168361-69168361 on Build GRCh38
rs536091898 Chr.5: 69168359-69168359 on Build GRCh38
rs754660428 Chr.5: 69168360-69168360 on Build GRCh38
rs767560839 Chr.5: 69168338-69168338 on Build GRCh38
rs750829238 Chr.5: 69168247-69168252 on Build GRCh38
rs759785076 Chr.5: 69168322-69168322 on Build GRCh38
rs768857099 Chr.5: 69168277-69168278 on Build GRCh38
rs370310551 Chr.5: 69168349-69168349 on Build GRCh38

View Full Product Details