Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3785674-3785923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPB
Entrez Gene ID
1059
Gene Name
centromere protein B
Gene Aliases
-
UniGene
Hs.516855
Gene Chromosome Location
Chr.20: 3783851-3786690 on Build GRCh38
Transcripts
NM_001810.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3693494 c.604G>A p.D202N Chr.20: 3785880-3785880 on Build GRCh38
6282114 c.622G>A p.A208T Chr.20: 3785862-3785862 on Build GRCh38
4885929 c.705C>A p.G235G Chr.20: 3785779-3785779 on Build GRCh38
7003354 c.732C>T p.A244A Chr.20: 3785752-3785752 on Build GRCh38
1307379 c.647G>A p.G216E Chr.20: 3785837-3785837 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758472789 Chr.20: 3785828-3785828 on Build GRCh38
rs369656252 Chr.20: 3785876-3785876 on Build GRCh38
rs767421155 Chr.20: 3785799-3785799 on Build GRCh38
rs748126240 Chr.20: 3785714-3785714 on Build GRCh38
rs879399143 Chr.20: 3785732-3785732 on Build GRCh38
rs767403202 Chr.20: 3785695-3785695 on Build GRCh38
rs770151730 Chr.20: 3785744-3785744 on Build GRCh38
rs777894487 Chr.20: 3785725-3785725 on Build GRCh38
rs776046699 Chr.20: 3785758-3785758 on Build GRCh38
rs760710349 Chr.20: 3785808-3785808 on Build GRCh38
rs897815576 Chr.20: 3785831-3785831 on Build GRCh38
rs1026671116 Chr.20: 3785717-3785717 on Build GRCh38
rs761497217 Chr.20: 3785797-3785797 on Build GRCh38
rs749791585 Chr.20: 3785903-3785903 on Build GRCh38
rs762412635 Chr.20: 3785764-3785764 on Build GRCh38
rs867769633 Chr.20: 3785697-3785697 on Build GRCh38
rs779076973 Chr.20: 3785707-3785707 on Build GRCh38
rs763658114 Chr.20: 3785771-3785771 on Build GRCh38
rs773929979 Chr.20: 3785781-3785781 on Build GRCh38
rs556461613 Chr.20: 3785861-3785861 on Build GRCh38
rs753775625 Chr.20: 3785822-3785822 on Build GRCh38
rs747225829 Chr.20: 3785727-3785727 on Build GRCh38
rs538468317 Chr.20: 3785770-3785770 on Build GRCh38
rs773929504 Chr.20: 3785906-3785906 on Build GRCh38
rs750199272 Chr.20: 3785806-3785806 on Build GRCh38
rs766276108 Chr.20: 3785813-3785813 on Build GRCh38
rs112241514 Chr.20: 3785745-3785745 on Build GRCh38
rs777853235 Chr.20: 3785846-3785846 on Build GRCh38
rs375303416 Chr.20: 3785743-3785743 on Build GRCh38
rs770168497 Chr.20: 3785887-3785887 on Build GRCh38
rs759724448 Chr.20: 3785866-3785866 on Build GRCh38
rs768060890 Chr.20: 3785904-3785904 on Build GRCh38
rs758481653 Chr.20: 3785720-3785720 on Build GRCh38
rs751786782 Chr.20: 3785860-3785860 on Build GRCh38
rs780496154 Chr.20: 3785890-3785890 on Build GRCh38
rs750397235 Chr.20: 3785698-3785698 on Build GRCh38
rs771299910 Chr.20: 3785733-3785733 on Build GRCh38

View Full Product Details