Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 67657981-67658252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYRK2
Entrez Gene ID
8445
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 2
Gene Aliases
-
UniGene
Hs.173135
Gene Chromosome Location
Chr.12: 67648732-67665405 on Build GRCh38
Transcripts
NM_003583.3 NM_006482.2 XM_017020032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
74471 c.1189G>T p.A397S Chr.12: 67658096-67658096 on Build GRCh38
2149676 c.1250C>A p.T417K Chr.12: 67658157-67658157 on Build GRCh38
4147415 c.1216T>G p.W406G Chr.12: 67658123-67658123 on Build GRCh38
271231 c.1155G>A p.S385S Chr.12: 67658062-67658062 on Build GRCh38
2099012 c.1124A>G p.E375G Chr.12: 67658031-67658031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374322116 Chr.12: 67658038-67658038 on Build GRCh38
rs755504526 Chr.12: 67658211-67658211 on Build GRCh38
rs981026091 Chr.12: 67658127-67658127 on Build GRCh38
rs749689088 Chr.12: 67658042-67658042 on Build GRCh38
rs745666311 Chr.12: 67658022-67658024 on Build GRCh38
rs749764319 Chr.12: 67658050-67658050 on Build GRCh38
rs909021101 Chr.12: 67658134-67658134 on Build GRCh38
rs56321998 Chr.12: 67658113-67658113 on Build GRCh38
rs368304778 Chr.12: 67658167-67658167 on Build GRCh38
rs766155721 Chr.12: 67658153-67658153 on Build GRCh38
rs143480472 Chr.12: 67658161-67658161 on Build GRCh38
rs146652644 Chr.12: 67658062-67658062 on Build GRCh38
rs140246773 Chr.12: 67658157-67658157 on Build GRCh38
rs757508984 Chr.12: 67658049-67658049 on Build GRCh38
rs780773474 Chr.12: 67658191-67658191 on Build GRCh38
rs756730510 Chr.12: 67658039-67658039 on Build GRCh38
rs144549473 Chr.12: 67658029-67658029 on Build GRCh38
rs886587503 Chr.12: 67658122-67658122 on Build GRCh38
rs768271747 Chr.12: 67658070-67658070 on Build GRCh38
rs369177246 Chr.12: 67658163-67658163 on Build GRCh38
rs754652492 Chr.12: 67658190-67658190 on Build GRCh38
rs918958302 Chr.12: 67658164-67658164 on Build GRCh38
rs747669756 Chr.12: 67658226-67658226 on Build GRCh38
rs543503984 Chr.12: 67658040-67658040 on Build GRCh38
rs779191268 Chr.12: 67658170-67658170 on Build GRCh38
rs561400923 Chr.12: 67658158-67658158 on Build GRCh38
rs376855807 Chr.12: 67658080-67658080 on Build GRCh38
rs772964517 Chr.12: 67658101-67658101 on Build GRCh38
rs769117431 Chr.12: 67658230-67658230 on Build GRCh38
rs769792355 Chr.12: 67658100-67658100 on Build GRCh38
rs148003575 Chr.12: 67658176-67658176 on Build GRCh38

View Full Product Details