Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 17069613-17069862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MICU3
Entrez Gene ID
286097
Gene Name
mitochondrial calcium uptake family member 3
Gene Aliases
EFHA2
UniGene
Hs.403594
Gene Chromosome Location
Chr.8: 17027238-17133069 on Build GRCh38
Transcripts
NM_181723.2 XM_005273477.2 XM_006716328.2 XM_006716329.2 XM_006716330.2 XM_006716333.3 XM_011544507.2 XM_011544508.2 XM_017013336.1 XM_017013337.1 XM_017013338.1 XM_017013339.1 XM_017013340.1 XM_017013341.1 XM_017013342.1 XM_017013343.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6411701 c.545C>T p.T182I Chr.8: 17069697-17069697 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754822698 Chr.8: 17069655-17069655 on Build GRCh38
rs888209180 Chr.8: 17069829-17069829 on Build GRCh38
rs867600533 Chr.8: 17069691-17069691 on Build GRCh38
rs563199959 Chr.8: 17069804-17069804 on Build GRCh38
rs755186716 Chr.8: 17069678-17069682 on Build GRCh38
rs772778483 Chr.8: 17069748-17069748 on Build GRCh38
rs369103171 Chr.8: 17069746-17069746 on Build GRCh38
rs776038204 Chr.8: 17069744-17069744 on Build GRCh38
rs756193526 Chr.8: 17069666-17069666 on Build GRCh38
rs144193351 Chr.8: 17069772-17069772 on Build GRCh38
rs762526712 Chr.8: 17069756-17069756 on Build GRCh38
rs370506546 Chr.8: 17069768-17069768 on Build GRCh38
rs955098837 Chr.8: 17069773-17069773 on Build GRCh38
rs751369104 Chr.8: 17069649-17069649 on Build GRCh38
rs372697343 Chr.8: 17069760-17069760 on Build GRCh38
rs779365540 Chr.8: 17069707-17069707 on Build GRCh38
rs929710786 Chr.8: 17069775-17069775 on Build GRCh38
rs372444696 Chr.8: 17069658-17069658 on Build GRCh38
rs376971575 Chr.8: 17069766-17069766 on Build GRCh38
rs17686794 Chr.8: 17069741-17069741 on Build GRCh38
rs542287414 Chr.8: 17069778-17069778 on Build GRCh38
rs1046780612 Chr.8: 17069792-17069792 on Build GRCh38
rs766040197 Chr.8: 17069758-17069758 on Build GRCh38
rs746268946 Chr.8: 17069735-17069735 on Build GRCh38
rs757554095 Chr.8: 17069704-17069704 on Build GRCh38
rs879403978 Chr.8: 17069652-17069652 on Build GRCh38
rs769333656 Chr.8: 17069747-17069747 on Build GRCh38
rs778053188 Chr.8: 17069677-17069677 on Build GRCh38
rs752616162 Chr.8: 17069659-17069659 on Build GRCh38
rs749515238 Chr.8: 17069701-17069701 on Build GRCh38

View Full Product Details