Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56365062-56365333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM210B
Entrez Gene ID
116151
Gene Name
family with sequence similarity 210 member B
Gene Aliases
5A3, C20orf108, dJ1167H4.1
UniGene
Hs.143736
Gene Chromosome Location
Chr.20: 56358927-56368663 on Build GRCh38
Transcripts
NM_080821.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
319064 c.205G>T p.G69W Chr.20: 56365105-56365105 on Build GRCh38
6004433 c.290G>A p.G97D Chr.20: 56365190-56365190 on Build GRCh38
3548020 c.187-1G>A p.? Chr.20: 56365086-56365086 on Build GRCh38
6242750 c.362+1_362+2insT p.? Chr.20: 56365263-56365264 on Build GRCh38
6389674 c.237G>T p.E79D Chr.20: 56365137-56365137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369855403 Chr.20: 56365291-56365291 on Build GRCh38
rs575667548 Chr.20: 56365130-56365130 on Build GRCh38
rs963746852 Chr.20: 56365287-56365287 on Build GRCh38
rs1029559781 Chr.20: 56365151-56365151 on Build GRCh38
rs940278056 Chr.20: 56365115-56365115 on Build GRCh38
rs371903531 Chr.20: 56365103-56365103 on Build GRCh38
rs776366938 Chr.20: 56365213-56365213 on Build GRCh38
rs755209154 Chr.20: 56365292-56365292 on Build GRCh38
rs781305698 Chr.20: 56365120-56365120 on Build GRCh38
rs889707700 Chr.20: 56365165-56365165 on Build GRCh38
rs768831283 Chr.20: 56365116-56365116 on Build GRCh38
rs144911122 Chr.20: 56365203-56365203 on Build GRCh38
rs757647256 Chr.20: 56365201-56365201 on Build GRCh38
rs780481691 Chr.20: 56365143-56365147 on Build GRCh38
rs141834554 Chr.20: 56365252-56365252 on Build GRCh38
rs538728588 Chr.20: 56365122-56365122 on Build GRCh38
rs575127516 Chr.20: 56365284-56365284 on Build GRCh38
rs763702536 Chr.20: 56365207-56365207 on Build GRCh38
rs17362970 Chr.20: 56365300-56365300 on Build GRCh38
rs774233187 Chr.20: 56365127-56365127 on Build GRCh38
rs530560365 Chr.20: 56365158-56365158 on Build GRCh38
rs756242005 Chr.20: 56365137-56365137 on Build GRCh38
rs748437483 Chr.20: 56365301-56365301 on Build GRCh38
rs148836702 Chr.20: 56365092-56365092 on Build GRCh38
rs149882892 Chr.20: 56365094-56365094 on Build GRCh38
rs780575478 Chr.20: 56365104-56365104 on Build GRCh38
rs775322634 Chr.20: 56365144-56365144 on Build GRCh38
rs764840904 Chr.20: 56365268-56365268 on Build GRCh38
rs558924794 Chr.20: 56365123-56365123 on Build GRCh38
rs200938298 Chr.20: 56365254-56365254 on Build GRCh38
rs771796777 Chr.20: 56365133-56365133 on Build GRCh38
rs375564926 Chr.20: 56365272-56365272 on Build GRCh38
rs555299952 Chr.20: 56365283-56365283 on Build GRCh38

View Full Product Details