Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 38351158-38351399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRED1
Entrez Gene ID
161742
Gene Name
sprouty related EVH1 domain containing 1
Gene Aliases
NFLS, PPP1R147, hSpred1, spred-1
UniGene
Hs.525781
Gene Chromosome Location
Chr.15: 38252087-38357249 on Build GRCh38
Transcripts
NM_152594.2 XM_005254202.3 XM_011521289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1227426 c.863A>C p.K288T Chr.15: 38351192-38351192 on Build GRCh38
7429387 c.869C>T p.S290L Chr.15: 38351198-38351198 on Build GRCh38
5849037 c.974G>A p.R325Q Chr.15: 38351303-38351303 on Build GRCh38
7153409 c.1047G>T p.R349S Chr.15: 38351376-38351376 on Build GRCh38
4054225 c.1028A>G p.N343S Chr.15: 38351357-38351357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899712805 Chr.15: 38351280-38351280 on Build GRCh38
rs535687467 Chr.15: 38351208-38351208 on Build GRCh38
rs886051105 Chr.15: 38351210-38351210 on Build GRCh38
rs750890621 Chr.15: 38351306-38351306 on Build GRCh38
rs765841849 Chr.15: 38351251-38351251 on Build GRCh38
rs140644874 Chr.15: 38351268-38351268 on Build GRCh38
rs146702985 Chr.15: 38351330-38351330 on Build GRCh38
rs761001792 Chr.15: 38351221-38351221 on Build GRCh38
rs375700452 Chr.15: 38351260-38351260 on Build GRCh38
rs115440602 Chr.15: 38351273-38351273 on Build GRCh38
rs749692794 Chr.15: 38351292-38351297 on Build GRCh38
rs369711772 Chr.15: 38351292-38351292 on Build GRCh38
rs554327738 Chr.15: 38351215-38351215 on Build GRCh38
rs752191960 Chr.15: 38351257-38351257 on Build GRCh38
rs764868020 Chr.15: 38351249-38351249 on Build GRCh38
rs201933663 Chr.15: 38351204-38351204 on Build GRCh38
rs1057517943 Chr.15: 38351377-38351389 on Build GRCh38
rs540124243 Chr.15: 38351228-38351228 on Build GRCh38
rs766814966 Chr.15: 38351267-38351267 on Build GRCh38
rs778423017 Chr.15: 38351195-38351203 on Build GRCh38
rs372665535 Chr.15: 38351195-38351195 on Build GRCh38
rs779966392 Chr.15: 38351324-38351324 on Build GRCh38
rs776909752 Chr.15: 38351235-38351235 on Build GRCh38
rs770375243 Chr.15: 38351364-38351364 on Build GRCh38
rs1014323023 Chr.15: 38351243-38351243 on Build GRCh38
rs765603808 Chr.15: 38351329-38351329 on Build GRCh38
rs1005383827 Chr.15: 38351270-38351270 on Build GRCh38
rs373477920 Chr.15: 38351335-38351335 on Build GRCh38
rs3751526 Chr.15: 38351373-38351373 on Build GRCh38
rs376679136 Chr.15: 38351360-38351360 on Build GRCh38
rs758102077 Chr.15: 38351323-38351323 on Build GRCh38
rs774835982 Chr.15: 38351374-38351374 on Build GRCh38
rs963408174 Chr.15: 38351353-38351353 on Build GRCh38
rs773833198 Chr.15: 38351366-38351366 on Build GRCh38
rs777379558 Chr.15: 38351193-38351193 on Build GRCh38
rs114636635 Chr.15: 38351255-38351255 on Build GRCh38
rs1028970466 Chr.15: 38351307-38351307 on Build GRCh38
rs756634323 Chr.15: 38351314-38351314 on Build GRCh38
rs200974459 Chr.15: 38351216-38351216 on Build GRCh38
rs753357510 Chr.15: 38351283-38351283 on Build GRCh38
rs987528002 Chr.15: 38351371-38351371 on Build GRCh38
rs768937237 Chr.15: 38351334-38351334 on Build GRCh38
rs1057518683 Chr.15: 38351302-38351302 on Build GRCh38
rs775556490 Chr.15: 38351217-38351217 on Build GRCh38

View Full Product Details