Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101761487-101761752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPTAB
Entrez Gene ID
79158
Gene Name
N-acetylglucosamine-1-phosphate transferase alpha and beta subunits
Gene Aliases
GNPTA, ICD
UniGene
Hs.46850
Gene Chromosome Location
Chr.12: 101745497-101830867 on Build GRCh38
Transcripts
NM_024312.4 XM_006719593.2 XM_011538731.2 XM_017019961.1 XM_017019962.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
199653 c.2824C>A p.L942I Chr.12: 101761655-101761655 on Build GRCh38
4038028 c.2805C>T p.L935L Chr.12: 101761674-101761674 on Build GRCh38
1358183 c.2775G>A p.R925R Chr.12: 101761704-101761704 on Build GRCh38
4038027 c.2884G>A p.D962N Chr.12: 101761595-101761595 on Build GRCh38
4807787 c.2851C>G p.R951G Chr.12: 101761628-101761628 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755785958 Chr.12: 101761620-101761620 on Build GRCh38
rs281865002 Chr.12: 101761702-101761702 on Build GRCh38
rs772991984 Chr.12: 101761559-101761559 on Build GRCh38
rs777346722 Chr.12: 101761627-101761627 on Build GRCh38
rs749088847 Chr.12: 101761628-101761628 on Build GRCh38
rs760807924 Chr.12: 101761582-101761582 on Build GRCh38
rs756617125 Chr.12: 101761650-101761650 on Build GRCh38
rs370081726 Chr.12: 101761551-101761551 on Build GRCh38
rs398124398 Chr.12: 101761583-101761583 on Build GRCh38
rs761868815 Chr.12: 101761709-101761709 on Build GRCh38
rs750799040 Chr.12: 101761595-101761595 on Build GRCh38
rs34260159 Chr.12: 101761617-101761618 on Build GRCh38
rs766838280 Chr.12: 101761599-101761599 on Build GRCh38
rs201970402 Chr.12: 101761549-101761549 on Build GRCh38
rs374361252 Chr.12: 101761566-101761566 on Build GRCh38
rs368650569 Chr.12: 101761593-101761593 on Build GRCh38
rs747616827 Chr.12: 101761618-101761618 on Build GRCh38
rs555336070 Chr.12: 101761575-101761575 on Build GRCh38
rs757574720 Chr.12: 101761721-101761722 on Build GRCh38
rs755493862 Chr.12: 101761610-101761610 on Build GRCh38
rs772153642 Chr.12: 101761667-101761667 on Build GRCh38
rs764229793 Chr.12: 101761588-101761588 on Build GRCh38
rs281865003 Chr.12: 101761696-101761696 on Build GRCh38
rs281865005 Chr.12: 101761612-101761612 on Build GRCh38
rs750905375 Chr.12: 101761629-101761629 on Build GRCh38
rs281865004 Chr.12: 101761613-101761613 on Build GRCh38
rs758750238 Chr.12: 101761598-101761598 on Build GRCh38
rs1020172077 Chr.12: 101761630-101761630 on Build GRCh38
rs111348341 Chr.12: 101761517-101761517 on Build GRCh38
rs762722597 Chr.12: 101761572-101761572 on Build GRCh38
rs776709484 Chr.12: 101761692-101761692 on Build GRCh38
rs752018430 Chr.12: 101761601-101761601 on Build GRCh38
rs192687061 Chr.12: 101761579-101761579 on Build GRCh38
rs745752214 Chr.12: 101761661-101761661 on Build GRCh38
rs765486254 Chr.12: 101761716-101761716 on Build GRCh38
rs762041904 Chr.12: 101761592-101761592 on Build GRCh38
rs138390866 Chr.12: 101761615-101761615 on Build GRCh38
rs780256662 Chr.12: 101761668-101761668 on Build GRCh38
rs774319874 Chr.12: 101761638-101761638 on Build GRCh38
rs561347282 Chr.12: 101761550-101761550 on Build GRCh38
rs997622954 Chr.12: 101761721-101761721 on Build GRCh38
rs768192010 Chr.12: 101761522-101761522 on Build GRCh38
rs773343494 Chr.12: 101761717-101761717 on Build GRCh38
rs780618202 Chr.12: 101761617-101761617 on Build GRCh38

View Full Product Details