Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32222501-32222726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3I
Entrez Gene ID
8668
Gene Name
eukaryotic translation initiation factor 3 subunit I
Gene Aliases
EIF3S2, PRO2242, TRIP-1, TRIP1, eIF3-beta, eIF3-p36
UniGene
Hs.530096
Gene Chromosome Location
Chr.1: 32221584-32231604 on Build GRCh38
Transcripts
NM_003757.3 XM_017002671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1687338 c.4A>G p.K2E Chr.1: 32222538-32222538 on Build GRCh38
414479 c.45G>C p.Q15H Chr.1: 32222579-32222579 on Build GRCh38
3966525 c.90G>A p.K30K Chr.1: 32222624-32222624 on Build GRCh38
70588 c.13C>G p.L5V Chr.1: 32222547-32222547 on Build GRCh38
358094 c.79A>T p.T27S Chr.1: 32222613-32222613 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745439479 Chr.1: 32222544-32222544 on Build GRCh38
rs377597986 Chr.1: 32222637-32222637 on Build GRCh38
rs760898883 Chr.1: 32222623-32222623 on Build GRCh38
rs769837607 Chr.1: 32222594-32222594 on Build GRCh38
rs368937076 Chr.1: 32222681-32222681 on Build GRCh38
rs754684442 Chr.1: 32222664-32222664 on Build GRCh38
rs961514513 Chr.1: 32222582-32222582 on Build GRCh38
rs775639469 Chr.1: 32222609-32222609 on Build GRCh38
rs774283623 Chr.1: 32222597-32222597 on Build GRCh38
rs776830433 Chr.1: 32222662-32222662 on Build GRCh38
rs552937919 Chr.1: 32222696-32222696 on Build GRCh38
rs551829220 Chr.1: 32222640-32222640 on Build GRCh38
rs11554223 Chr.1: 32222570-32222570 on Build GRCh38
rs759710484 Chr.1: 32222602-32222602 on Build GRCh38
rs753038401 Chr.1: 32222523-32222523 on Build GRCh38
rs748035113 Chr.1: 32222677-32222677 on Build GRCh38
rs201570798 Chr.1: 32222535-32222535 on Build GRCh38
rs765817885 Chr.1: 32222656-32222656 on Build GRCh38
rs749865587 Chr.1: 32222661-32222661 on Build GRCh38
rs757689383 Chr.1: 32222652-32222652 on Build GRCh38
rs201980498 Chr.1: 32222530-32222530 on Build GRCh38
rs746693785 Chr.1: 32222567-32222567 on Build GRCh38
rs971361857 Chr.1: 32222621-32222621 on Build GRCh38
rs201494062 Chr.1: 32222673-32222673 on Build GRCh38
rs780925879 Chr.1: 32222655-32222655 on Build GRCh38
rs768527049 Chr.1: 32222571-32222571 on Build GRCh38
rs747972544 Chr.1: 32222589-32222589 on Build GRCh38
rs756429807 Chr.1: 32222524-32222524 on Build GRCh38
rs746604002 Chr.1: 32222663-32222663 on Build GRCh38
rs140164523 Chr.1: 32222587-32222587 on Build GRCh38
rs779407248 Chr.1: 32222691-32222691 on Build GRCh38
rs1054218575 Chr.1: 32222689-32222689 on Build GRCh38
rs779888450 Chr.1: 32222564-32222564 on Build GRCh38
rs945169174 Chr.1: 32222707-32222707 on Build GRCh38
rs754532538 Chr.1: 32222597-32222598 on Build GRCh38
rs141934338 Chr.1: 32222561-32222561 on Build GRCh38
rs778279137 Chr.1: 32222539-32222539 on Build GRCh38
rs778133280 Chr.1: 32222525-32222525 on Build GRCh38
rs536664845 Chr.1: 32222603-32222603 on Build GRCh38

View Full Product Details