Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 16212392-16212647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT15
Entrez Gene ID
117248
Gene Name
polypeptide N-acetylgalactosaminyltransferase 15
Gene Aliases
GALNACT15, GALNT13, GALNT7, GALNTL2, PIH5, pp-GalNAc-T15
UniGene
Hs.411308
Gene Chromosome Location
Chr.3: 16174322-16245190 on Build GRCh38
Transcripts
NM_001319051.1 NM_054110.4 XM_005264852.4 XM_017005682.1 XM_017005683.1 XM_017005684.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7172053 c.1231C>A p.P411T Chr.3: 16212602-16212602 on Build GRCh38
396026 c.1198-1G>T p.? Chr.3: 16212568-16212568 on Build GRCh38
4598219 c.1244delT p.V415fs*20 Chr.3: 16212615-16212615 on Build GRCh38
6096682 c.1267G>T p.D423Y Chr.3: 16212638-16212638 on Build GRCh38
3373071 c.1251C>T p.H417H Chr.3: 16212622-16212622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769184724 Chr.3: 16212614-16212614 on Build GRCh38
rs748067913 Chr.3: 16212516-16212516 on Build GRCh38
rs143119163 Chr.3: 16212517-16212517 on Build GRCh38
rs749457469 Chr.3: 16212553-16212553 on Build GRCh38
rs941404278 Chr.3: 16212478-16212478 on Build GRCh38
rs772839615 Chr.3: 16212620-16212620 on Build GRCh38
rs571482279 Chr.3: 16212439-16212439 on Build GRCh38
rs762736653 Chr.3: 16212527-16212527 on Build GRCh38
rs761852083 Chr.3: 16212612-16212612 on Build GRCh38
rs182370588 Chr.3: 16212451-16212451 on Build GRCh38
rs570209017 Chr.3: 16212496-16212496 on Build GRCh38
rs766719585 Chr.3: 16212530-16212530 on Build GRCh38
rs777973052 Chr.3: 16212545-16212545 on Build GRCh38
rs771289364 Chr.3: 16212561-16212561 on Build GRCh38
rs752153411 Chr.3: 16212603-16212603 on Build GRCh38
rs779927324 Chr.3: 16212570-16212570 on Build GRCh38
rs894813344 Chr.3: 16212587-16212587 on Build GRCh38
rs756114418 Chr.3: 16212538-16212538 on Build GRCh38
rs1014017140 Chr.3: 16212420-16212420 on Build GRCh38
rs759084722 Chr.3: 16212514-16212514 on Build GRCh38
rs34031379 Chr.3: 16212465-16212465 on Build GRCh38
rs781677334 Chr.3: 16212537-16212537 on Build GRCh38
rs750014608 Chr.3: 16212525-16212525 on Build GRCh38
rs955474548 Chr.3: 16212422-16212422 on Build GRCh38
rs528413099 Chr.3: 16212416-16212416 on Build GRCh38
rs1027594998 Chr.3: 16212502-16212502 on Build GRCh38
rs746830716 Chr.3: 16212590-16212590 on Build GRCh38
rs369561893 Chr.3: 16212536-16212536 on Build GRCh38
rs776688286 Chr.3: 16212611-16212611 on Build GRCh38
rs1038598362 Chr.3: 16212510-16212510 on Build GRCh38
rs551406572 Chr.3: 16212421-16212421 on Build GRCh38
rs961770094 Chr.3: 16212454-16212454 on Build GRCh38

View Full Product Details