Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 37841715-37841944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNLT3
Entrez Gene ID
6990
Gene Name
dynein light chain Tctex-type 3
Gene Aliases
RP3, TCTE1L, TCTEX1L
UniGene
Hs.446392
Gene Chromosome Location
Chr.X: 37838836-37847636 on Build GRCh38
Transcripts
NM_006520.2

SNP Detail

SNP ID Chromosome Location
rs770416498 Chr.X: 37841915-37841915 on Build GRCh38
rs759653980 Chr.X: 37841860-37841862 on Build GRCh38
rs375297123 Chr.X: 37841755-37841755 on Build GRCh38
rs778559587 Chr.X: 37841890-37841890 on Build GRCh38
rs764059172 Chr.X: 37841867-37841867 on Build GRCh38
rs760584132 Chr.X: 37841820-37841820 on Build GRCh38
rs138075306 Chr.X: 37841792-37841792 on Build GRCh38
rs200958514 Chr.X: 37841895-37841895 on Build GRCh38
rs756778054 Chr.X: 37841881-37841881 on Build GRCh38
rs371256878 Chr.X: 37841770-37841770 on Build GRCh38
rs748686635 Chr.X: 37841752-37841752 on Build GRCh38
rs767250789 Chr.X: 37841806-37841806 on Build GRCh38
rs750073352 Chr.X: 37841894-37841894 on Build GRCh38
rs927537903 Chr.X: 37841756-37841756 on Build GRCh38
rs781673184 Chr.X: 37841896-37841896 on Build GRCh38
rs776670879 Chr.X: 37841810-37841810 on Build GRCh38
rs753343380 Chr.X: 37841879-37841879 on Build GRCh38
rs765407488 Chr.X: 37841887-37841905 on Build GRCh38
rs11544207 Chr.X: 37841854-37841854 on Build GRCh38
rs188876827 Chr.X: 37841913-37841913 on Build GRCh38
rs144229104 Chr.X: 37841815-37841815 on Build GRCh38
rs11771 Chr.X: 37841795-37841795 on Build GRCh38
rs771169855 Chr.X: 37841782-37841782 on Build GRCh38
rs763189332 Chr.X: 37841779-37841779 on Build GRCh38

View Full Product Details