Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65020340-65020595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL2
Entrez Gene ID
402
Gene Name
ADP ribosylation factor like GTPase 2
Gene Aliases
ARFL2
UniGene
Hs.502836
Gene Chromosome Location
Chr.11: 65014113-65022185 on Build GRCh38
Transcripts
NM_001199745.1 NM_001667.3 NR_037650.1

Gene Symbol
ARL2-SNX15
Entrez Gene ID
100528018
Gene Name
ARL2-SNX15 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.80132
Gene Chromosome Location
Chr.11: 65014113-65040572 on Build GRCh38
Transcripts
NM_001199745.1 NM_001667.3 NR_037650.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6604808 c.397C>G p.L133V Chr.11: 65020476-65020476 on Build GRCh38
3782660 c.401C>T p.S134F Chr.11: 65020480-65020480 on Build GRCh38
1355859 c.409G>A p.A137T Chr.11: 65020488-65020488 on Build GRCh38
6134068 c.376A>G p.K126E Chr.11: 65020455-65020455 on Build GRCh38
6552252 c.349G>T p.G117* Chr.11: 65020428-65020428 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576735088 Chr.11: 65020391-65020391 on Build GRCh38
rs560909889 Chr.11: 65020525-65020525 on Build GRCh38
rs774959134 Chr.11: 65020426-65020426 on Build GRCh38
rs367666014 Chr.11: 65020489-65020489 on Build GRCh38
rs746418004 Chr.11: 65020495-65020495 on Build GRCh38
rs140716291 Chr.11: 65020428-65020428 on Build GRCh38
rs946607464 Chr.11: 65020431-65020431 on Build GRCh38
rs762322883 Chr.11: 65020442-65020442 on Build GRCh38
rs780202680 Chr.11: 65020458-65020458 on Build GRCh38
rs777574623 Chr.11: 65020395-65020395 on Build GRCh38
rs750976395 Chr.11: 65020454-65020454 on Build GRCh38
rs370382116 Chr.11: 65020439-65020439 on Build GRCh38
rs145536227 Chr.11: 65020494-65020494 on Build GRCh38
rs924304189 Chr.11: 65020427-65020427 on Build GRCh38
rs150084832 Chr.11: 65020463-65020463 on Build GRCh38
rs768041336 Chr.11: 65020522-65020522 on Build GRCh38
rs779164260 Chr.11: 65020473-65020473 on Build GRCh38
rs200325022 Chr.11: 65020397-65020397 on Build GRCh38
rs992965526 Chr.11: 65020435-65020435 on Build GRCh38
rs745842096 Chr.11: 65020493-65020493 on Build GRCh38
rs756124622 Chr.11: 65020457-65020457 on Build GRCh38
rs746804914 Chr.11: 65020479-65020479 on Build GRCh38
rs762233432 Chr.11: 65020515-65020515 on Build GRCh38
rs756281517 Chr.11: 65020388-65020388 on Build GRCh38
rs770754216 Chr.11: 65020487-65020487 on Build GRCh38
rs764491058 Chr.11: 65020430-65020430 on Build GRCh38
rs79393344 Chr.11: 65020408-65020408 on Build GRCh38
rs373752729 Chr.11: 65020394-65020394 on Build GRCh38
rs966051759 Chr.11: 65020366-65020366 on Build GRCh38
rs34747203 Chr.11: 65020559-65020559 on Build GRCh38
rs769398732 Chr.11: 65020425-65020425 on Build GRCh38
rs770842028 Chr.11: 65020400-65020400 on Build GRCh38
rs768405201 Chr.11: 65020497-65020497 on Build GRCh38
rs965201198 Chr.11: 65020361-65020361 on Build GRCh38
rs774211097 Chr.11: 65020514-65020514 on Build GRCh38
rs750453805 Chr.11: 65020387-65020387 on Build GRCh38
rs749571711 Chr.11: 65020393-65020393 on Build GRCh38
rs374080193 Chr.11: 65020464-65020464 on Build GRCh38
rs776618009 Chr.11: 65020406-65020406 on Build GRCh38
rs768131237 Chr.11: 65020449-65020449 on Build GRCh38
rs368433913 Chr.11: 65020526-65020526 on Build GRCh38
rs781358640 Chr.11: 65020372-65020372 on Build GRCh38
rs762451444 Chr.11: 65020496-65020496 on Build GRCh38

View Full Product Details