Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 29307439-29307710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAR2
Entrez Gene ID
55711
Gene Name
fatty acyl-CoA reductase 2
Gene Aliases
HEL-S-81, MLSTD1, SDR10E2
UniGene
Hs.684482
Gene Chromosome Location
Chr.12: 29149003-29335616 on Build GRCh38
Transcripts
NM_001271783.1 NM_001271784.1 NM_018099.4 XM_011520747.2 XM_011520748.2 XM_017019624.1 XM_017019625.1 NR_103860.1

Gene Symbol
LOC100506606
Entrez Gene ID
100506606
Gene Name
uncharacterized LOC100506606
Gene Aliases
-
UniGene
Hs.298851
Gene Chromosome Location
Chr.12: 29280415-29317848 on Build GRCh38
Transcripts
NM_001271783.1 NM_001271784.1 NM_018099.4 XM_011520747.2 XM_011520748.2 XM_017019624.1 XM_017019625.1 NR_103860.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4552231 c.553G>A p.D185N Chr.12: 29307665-29307665 on Build GRCh38
4963314 c.573G>T p.E191D Chr.12: 29307685-29307685 on Build GRCh38
7298379 c.564T>C p.I188I Chr.12: 29307676-29307676 on Build GRCh38
3376228 c.570C>T p.D190D Chr.12: 29307682-29307682 on Build GRCh38
5481026 c.576T>C p.I192I Chr.12: 29307688-29307688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs947890386 Chr.12: 29307585-29307585 on Build GRCh38
rs903250514 Chr.12: 29307631-29307631 on Build GRCh38
rs188101812 Chr.12: 29307537-29307537 on Build GRCh38
rs374567986 Chr.12: 29307556-29307556 on Build GRCh38
rs769914306 Chr.12: 29307622-29307623 on Build GRCh38
rs144553899 Chr.12: 29307682-29307682 on Build GRCh38
rs771863489 Chr.12: 29307620-29307620 on Build GRCh38
rs774825418 Chr.12: 29307667-29307667 on Build GRCh38
rs919026032 Chr.12: 29307496-29307496 on Build GRCh38
rs115529954 Chr.12: 29307490-29307490 on Build GRCh38
rs1048890249 Chr.12: 29307601-29307601 on Build GRCh38
rs912060472 Chr.12: 29307465-29307465 on Build GRCh38
rs916473644 Chr.12: 29307575-29307575 on Build GRCh38
rs777241030 Chr.12: 29307645-29307645 on Build GRCh38
rs568929138 Chr.12: 29307511-29307511 on Build GRCh38
rs765318411 Chr.12: 29307655-29307655 on Build GRCh38
rs142002128 Chr.12: 29307643-29307647 on Build GRCh38
rs944892479 Chr.12: 29307482-29307482 on Build GRCh38
rs573237051 Chr.12: 29307603-29307603 on Build GRCh38
rs75909823 Chr.12: 29307476-29307476 on Build GRCh38
rs748912572 Chr.12: 29307613-29307613 on Build GRCh38
rs953395971 Chr.12: 29307683-29307683 on Build GRCh38
rs747735367 Chr.12: 29307616-29307616 on Build GRCh38
rs767815413 Chr.12: 29307475-29307476 on Build GRCh38
rs773698282 Chr.12: 29307622-29307622 on Build GRCh38
rs2288125 Chr.12: 29307615-29307615 on Build GRCh38
rs937928943 Chr.12: 29307576-29307576 on Build GRCh38
rs866552711 Chr.12: 29307555-29307555 on Build GRCh38
rs377282784 Chr.12: 29307626-29307626 on Build GRCh38
rs766761015 Chr.12: 29307634-29307634 on Build GRCh38
rs1056312680 Chr.12: 29307581-29307581 on Build GRCh38
rs977431112 Chr.12: 29307483-29307483 on Build GRCh38
rs115785862 Chr.12: 29307567-29307567 on Build GRCh38
rs555974387 Chr.12: 29307649-29307649 on Build GRCh38

View Full Product Details