Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133385526-133385799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STKLD1
Entrez Gene ID
169436
Gene Name
serine/threonine kinase like domain containing 1
Gene Aliases
C9orf96, SgK071, Sk521
UniGene
Hs.720195
Gene Chromosome Location
Chr.9: 133376413-133406096 on Build GRCh38
Transcripts
NM_153710.4 NR_103997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6114921 c.222G>A p.L74L Chr.9: 133385619-133385619 on Build GRCh38
4138880 c.294+7C>A p.? Chr.9: 133385698-133385698 on Build GRCh38
6013680 c.242G>A p.R81Q Chr.9: 133385639-133385639 on Build GRCh38
5826485 c.241C>T p.R81W Chr.9: 133385638-133385638 on Build GRCh38
6240044 c.267G>T p.Q89H Chr.9: 133385664-133385664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782744859 Chr.9: 133385663-133385663 on Build GRCh38
rs781895173 Chr.9: 133385686-133385686 on Build GRCh38
rs782619978 Chr.9: 133385689-133385689 on Build GRCh38
rs782252466 Chr.9: 133385697-133385697 on Build GRCh38
rs781872920 Chr.9: 133385570-133385570 on Build GRCh38
rs992806126 Chr.9: 133385776-133385776 on Build GRCh38
rs371788326 Chr.9: 133385578-133385578 on Build GRCh38
rs782372331 Chr.9: 133385623-133385623 on Build GRCh38
rs199937057 Chr.9: 133385592-133385592 on Build GRCh38
rs781824940 Chr.9: 133385569-133385569 on Build GRCh38
rs375163386 Chr.9: 133385587-133385587 on Build GRCh38
rs369047700 Chr.9: 133385643-133385643 on Build GRCh38
rs587673686 Chr.9: 133385728-133385728 on Build GRCh38
rs189765356 Chr.9: 133385716-133385716 on Build GRCh38
rs782395874 Chr.9: 133385729-133385729 on Build GRCh38
rs781992947 Chr.9: 133385627-133385627 on Build GRCh38
rs782806461 Chr.9: 133385646-133385646 on Build GRCh38
rs781931381 Chr.9: 133385694-133385694 on Build GRCh38
rs1044518049 Chr.9: 133385556-133385556 on Build GRCh38
rs587603094 Chr.9: 133385767-133385767 on Build GRCh38
rs587731241 Chr.9: 133385758-133385758 on Build GRCh38
rs781858465 Chr.9: 133385654-133385654 on Build GRCh38
rs782017435 Chr.9: 133385724-133385724 on Build GRCh38
rs117173599 Chr.9: 133385713-133385713 on Build GRCh38
rs913232213 Chr.9: 133385741-133385741 on Build GRCh38
rs34399743 Chr.9: 133385574-133385574 on Build GRCh38
rs782359483 Chr.9: 133385639-133385639 on Build GRCh38
rs978371710 Chr.9: 133385768-133385768 on Build GRCh38
rs782327061 Chr.9: 133385723-133385723 on Build GRCh38
rs782625529 Chr.9: 133385714-133385714 on Build GRCh38
rs782766566 Chr.9: 133385691-133385691 on Build GRCh38
rs781850148 Chr.9: 133385667-133385667 on Build GRCh38
rs145815775 Chr.9: 133385679-133385679 on Build GRCh38
rs956328491 Chr.9: 133385774-133385774 on Build GRCh38
rs369981983 Chr.9: 133385571-133385571 on Build GRCh38
rs924714550 Chr.9: 133385770-133385770 on Build GRCh38
rs782129776 Chr.9: 133385661-133385661 on Build GRCh38
rs371581139 Chr.9: 133385593-133385593 on Build GRCh38
rs781952636 Chr.9: 133385733-133385733 on Build GRCh38
rs782548825 Chr.9: 133385687-133385687 on Build GRCh38
rs150285987 Chr.9: 133385678-133385678 on Build GRCh38
rs782138835 Chr.9: 133385591-133385591 on Build GRCh38
rs992066701 Chr.9: 133385668-133385668 on Build GRCh38
rs782064279 Chr.9: 133385644-133385644 on Build GRCh38
rs990133892 Chr.9: 133385737-133385737 on Build GRCh38
rs375343281 Chr.9: 133385638-133385638 on Build GRCh38
rs1008554399 Chr.9: 133385608-133385608 on Build GRCh38

View Full Product Details