Gene Transcripts

Gene Symbol
Abcg1
Entrez Gene ID
11307
Gene Name
ATP-binding cassette, sub-family G (WHITE), member 1
Gene Aliases
AW413978, Abc8, White
Chromosome Location
Chr. 17: 31057694 - 31117984 on Build GRCm38
UniGene ID
Mm.15691
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009593.2 NP_033723.1 8 1027
GenBank mRNA AF323659.1 - 8 1023
BC119471.2 - 8 975 40048166
U34920.1 - 8 985
Z48745.1 - 8 1014

View Full Product Details