Gene Transcripts

Gene Symbol
Ovgp1
Entrez Gene ID
12659
Gene Name
oviductal glycoprotein 1
Gene Aliases
120kDa, AU016433, AU019448, Chit5, MOGP, OGP, muc9
Chromosome Location
Chr. 3: 105973802 - 105987423 on Build GRCm38
UniGene ID
Mm.431947
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007696.2 NP_031722.1 8 828
GenBank mRNA AK054141.1 - 8 829
AY521455.1 - 8 814
BC137995.1 - 8 919 8861014
D32137.1 - 8 828

Gene Symbol
Wdr77
Entrez Gene ID
70465
Gene Name
WD repeat domain 77
Gene Aliases
2610003I18Rik, 2610312E17Rik, C79984, p44/MEP50
Chromosome Location
Chr. 3: 105959498 - 105969760 on Build GRCm38
UniGene ID
Mm.5110
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK163839.1 - 2332

View Full Product Details