Gene Transcripts

Gene Symbol
Fanca
Entrez Gene ID
14087
Gene Name
Fanconi anemia, complementation group A
Gene Aliases
AW208693, FACA
Chromosome Location
Chr. 8: 123268244 - 123318651 on Build GRCm38
UniGene ID
Mm.444516
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016925.3 NP_058621.2 10 870
GenBank mRNA AF208116.1 - 10 872
AF208117.1 - 7 553
AF208118.1 - 10 872
AK084990.1 - 9 978
AK166112.1 - 10 855
BC150705.1 - 10 868 9087616

View Full Product Details