Gene Transcripts

Gene Symbol
Sin3a
Entrez Gene ID
20466
Gene Name
transcriptional regulator, SIN3A (yeast)
Gene Aliases
AW553200, Sin3, mKIAA4126, mSin3A
Chromosome Location
Chr. 9: 57071991 - 57128368 on Build GRCm38
UniGene ID
Mm.15755
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110350.1 NP_001103820.1 15 2932
NM_001110351.1 NP_001103821.1 16 2809
NM_011378.2 NP_035508.2 16 3040
GenBank mRNA AK032209.1 - 15 2800
AK136347.1 - 10 1734
AK136972.1 - 1 165
AK220292.1 - 15 2909
BC052716.1 - 15 2819 6837144
BC053385.1 - 15 2802
BC078454.1 - 15 2793
L36831.1 - 15 2723
L38620.2 - 14 2699
L38621.2 - 14 2699
U22394.1 - 16 3048

View Full Product Details