Gene Transcripts

Gene Symbol
Arhgap25
Entrez Gene ID
232201
Gene Name
Rho GTPase activating protein 25
Gene Aliases
A130039I20Rik
Chromosome Location
Chr. 6: 87458544 - 87533259 on Build GRCm38
UniGene ID
Mm.119564
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037727.2 NP_001032816.1 10 1874
NM_001286610.1 NP_001273539.1 9 1674
NM_175476.4 NP_780685.2 10 1545
GenBank mRNA AK037710.1 - 10 1607
AK048450.1 - 10 1505
AK137444.1 - 10 1533
AK137526.1 - 10 1945
AK159598.1 - 9 1675
AK159941.1 - 9 1675
AK172878.1 - 8 1204
BC108400.1 - 7 1155
BC138751.1 - 9 1453 8861773
BC138752.1 - 9 1453 8861774

View Full Product Details