Gene Transcripts

Gene Symbol
Vmn2r5
Entrez Gene ID
667060
Gene Name
vomeronasal 2, receptor 5
Gene Aliases
EG667060
Chromosome Location
Chr. 3: 64490783 - 64534962 on Build GRCm38
UniGene ID
Mm.484957
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104618.1 NP_001098088.1 3 1342

Gene Symbol
Vmn2r-ps11
Entrez Gene ID
319210
Gene Name
vomeronasal 2, receptor, pseudogene 11
Gene Aliases
4930518C23Rik
Chromosome Location
Chr. 3: 64632863 - 64844743 on Build GRCm38
UniGene ID
Mm.246646
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK029734.1 - 6 1709

View Full Product Details