Gene Transcripts

Gene Symbol
Dctn3
Entrez Gene ID
53598
Gene Name
dynactin 3
Gene Aliases
p24
Chromosome Location
Chr. 4: 41714798 - 41723163 on Build GRCm38
UniGene ID
Mm.29025
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159565.1 NP_001153037.1 4 333
NM_016890.4 NP_058586.3 4 333
GenBank mRNA AF098508.1 - 4 312
AK002720.1 - 4 324
AK003255.1 - 4 308
AK011840.1 - 4 330
AK159344.1 - 4 331
AV493721.1 - 4 319
BC061120.1 - 4 314 6774742

View Full Product Details