Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020330.4 | NP_065063.1 | 2 | 2630 | |
GenBank mRNA | AF251559.1 | - | 1 | 2122 | |
AK014827.2 | - | 2 | 2631 | ||
BC132152.1 | - | 1 | 2173 | 40130429 | |
BC132344.1 | - | 1 | 2173 | 40130621 |