Gene Transcripts
- Gene Symbol
- Dnaja4
- Entrez Gene ID
- 58233
- Gene Name
- DnaJ (Hsp40) homolog, subfamily A, member 4
- Gene Aliases
- 1110021L12Rik, AV358213, Dj4, Hsj4, mmDjA4
- Chromosome Location
- Chr. 9: 54696590 - 54716317 on Build GRCm38
- UniGene ID
- Mm.489676
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021422.4 |
NP_067397.1
|
5, 6 |
1046 |
|
GenBank mRNA |
AB032401.1 |
- |
6, 7 |
913 |
|
AK003903.1 |
- |
1, 2 |
187 |
|
AK016666.1 |
- |
4, 5 |
643 |
|
AK046476.1 |
- |
5, 6 |
1047 |
|
AK076175.1 |
- |
5, 6 |
1047 |
|
AK141909.1 |
- |
5, 6 |
1048 |
|
BC022948.1 |
- |
3, 4 |
544 |
|
BC147484.1 |
- |
5, 6 |
894 |
9056231 |
BC147486.1 |
- |
5, 6 |
894 |
9056239 |
View Full Product Details