Gene Transcripts

Gene Symbol
Erlec1
Entrez Gene ID
66753
Gene Name
endoplasmic reticulum lectin 1
Gene Aliases
4933407N01Rik
Chromosome Location
Chr. 11: 30865632 - 30954173 on Build GRCm38
UniGene ID
Mm.294641
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025745.3 NP_080021.3 7 652
GenBank mRNA AK002471.1 - 7 643
AK077120.1 - 7 645
AK133874.1 - 7 653
BC018468.1 - 7 662 3591901

View Full Product Details