Gene Transcripts

Gene Symbol
Clybl
Entrez Gene ID
69634
Gene Name
citrate lyase beta like
Gene Aliases
0610033J05Rik, 2310014M14Rik, AI256068, Clb
Chromosome Location
Chr. 14: 122171283 - 122402234 on Build GRCm38
UniGene ID
Mm.34608
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029556.3 NP_083832.2 5 615
GenBank mRNA AF428254.1 - 5 588
AK002741.1 - 5 742
AK009345.1 - 5 603
BC023398.1 - 5 605 5064709

View Full Product Details