Gene Transcripts

Gene Symbol
1700057G04Rik
Entrez Gene ID
78459
Gene Name
RIKEN cDNA 1700057G04 gene
Gene Aliases
Chromosome Location
Chr. 9: 92309327 - 92357876 on Build GRCm38
UniGene ID
Mm.179195
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033184.3 NP_001028356.1 5 582
GenBank mRNA AK132905.1 - 5 583
BC147543.1 - 4 472 9056534
BC172079.1 - 3 429

View Full Product Details