Gene Transcripts

Gene Symbol
Zfp3
Entrez Gene ID
193043
Gene Name
zinc finger protein 3
Gene Aliases
AI854635, B230309J24, Fnp-1, Zfp-3
Chromosome Location
Chr. 11: 70764447 - 70772928 on Build GRCm38
UniGene ID
Mm.40966
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177565.3 NP_808233.1 2 251
GenBank mRNA AK045753.1 - 2 252
BC096463.1 - 2 233 6466366

View Full Product Details