Gene Transcripts

Gene Symbol
Olfr342
Entrez Gene ID
258950
Gene Name
olfactory receptor 342
Gene Aliases
MOR136-4
Chromosome Location
Chr. 2: 36527414 - 36528352 on Build GRCm38
UniGene ID
Mm.223345
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146948.1 NP_667159.1 1 484
GenBank mRNA BC120521.1 - 1 510
BC120523.1 - 1 510 40129446

View Full Product Details