Gene Transcripts

Gene Symbol
Qdpr
Entrez Gene ID
110391
Gene Name
quinoid dihydropteridine reductase
Gene Aliases
2610008L04Rik, D5Ertd371e, Dhpr, PKU2
Chromosome Location
Chr. 5: 45434032 - 45450229 on Build GRCm38
UniGene ID
Mm.30204
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024236.2 NP_077198.1 7 798
GenBank mRNA AK011351.1 - 7 727
AK078123.1 - 7 751
AK159107.1 - 7 742
AK161660.1 - 7 691
AK166694.1 - 7 752
BC002107.1 - 7 738 3487104

View Full Product Details