Gene Transcripts

Gene Symbol
Gzmc
Entrez Gene ID
14940
Gene Name
granzyme C
Gene Aliases
AI323531, CCP2, Ctla-5, Ctla5
Chromosome Location
Chr. 14: 56231397 - 56241639 on Build GRCm38
UniGene ID
Mm.14465
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010371.2 NP_034501.2 2 185
GenBank mRNA BC120755.1 - 2 135
BC120757.1 - 2 135 40129680
BC145030.1 - 2 135 9053578
M18459.1 - 2 185
X12822.1 - 2 129

View Full Product Details