Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161767.1 | NP_001155239.1 | 12 | 2245 | |
NM_001161768.1 | NP_001155240.1 | 12 | 2242 | ||
NM_172451.3 | NP_766039.2 | 12 | 2241 | ||
GenBank mRNA | AK028506.1 | - | 12 | 2241 | |
AK049222.1 | - | 12 | 2244 | ||
AK155008.1 | - | 12 | 2248 | ||
AK159721.1 | - | 12 | 2223 | ||
BC119324.1 | - | 10 | 1894 | 8734073 | |
BC119326.1 | - | 10 | 1894 | 8734075 |