Gene Transcripts

Gene Symbol
Gm15800
Entrez Gene ID
269700
Gene Name
predicted gene 15800
Gene Aliases
A130010L24, AU042671, BC019639, EG545802, OTTMUSG00000026283, mKIAA0614
Chromosome Location
Chr. 5: 121219339 - 121368577 on Build GRCm38
UniGene ID
Mm.184589
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181421.4 NP_852086.3 63 11276
GenBank mRNA AK037367.1 - 2 253
AK129172.1 - 3 1248
AK139096.1 - 3 994
BC058664.1 - 3 955

View Full Product Details