Gene Transcripts

Gene Symbol
Zfp236
Entrez Gene ID
329002
Gene Name
zinc finger protein 236
Gene Aliases
AI447957, Gm335
Chromosome Location
Chr. 18: 82593593 - 82693627 on Build GRCm38
UniGene ID
Mm.34438
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177832.3 NP_808500.2 4 560
GenBank mRNA BC138187.1 - 4 560
BC145179.1 - 4 560
BC145180.1 - 4 560

View Full Product Details