Gene Transcripts

Gene Symbol
Ly6g6c
Entrez Gene ID
68468
Gene Name
lymphocyte antigen 6 complex, locus G6C
Gene Aliases
1110003M04Rik, AU016360, G6c, NG24
Chromosome Location
Chr. 17: 35067325 - 35070048 on Build GRCm38
UniGene ID
Mm.215096
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023463.3 NP_075952.1 2 133
GenBank mRNA AJ315546.1 - 2 68
AJ315547.1 - 2 68
AK003375.1 - 2 134
BC116366.1 - 2 97 40100049

View Full Product Details