Gene Transcripts

Gene Symbol
2310079G19Rik
Entrez Gene ID
69699
Gene Name
RIKEN cDNA 2310079G19 gene
Gene Aliases
Chromosome Location
Chr. 16: 88626788 - 88627666 on Build GRCm38
UniGene ID
Mm.158296
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027173.1 NP_081449.1 1 579
GenBank mRNA AK010217.1 - 1 581

View Full Product Details