Gene Transcripts

Gene Symbol
Jsrp1
Entrez Gene ID
71912
Gene Name
junctional sarcoplasmic reticulum protein 1
Gene Aliases
2300003C06Rik, 2310032K21Rik, JP-45, JP45
Chromosome Location
Chr. 10: 80808494 - 80813593 on Build GRCm38
UniGene ID
Mm.45137
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028001.3 NP_082277.2 7 835
GenBank mRNA AK009016.1 - 7 829
AK009578.1 - 7 1050
BC104136.1 - 6 749
BC104137.1 - 6 750

View Full Product Details