Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145824.4 | NP_665823.2 | 13 | 1834 | |
GenBank mRNA | AK087165.1 | - | 3 | 251 | |
AK173177.1 | - | 13 | 1647 | ||
AY337314.1 | - | 13 | 1835 | ||
BC024698.1 | - | 13 | 1679 | 3667000 | |
EU281316.1 | - | 13 | 1695 |