Gene Transcripts

Gene Symbol
Wdr44
Entrez Gene ID
246152
Gene Name
WD repeat domain 44
Gene Aliases
Chromosome Location
Chr. X: 120860003 - 120965918 on Build Rnor_6.0
UniGene ID
Rn.79380
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100823.1 NP_001094293.1 13 2059
XM_006257442.2 XP_006257504.1 13 2246
GenBank mRNA AF130121.1 - 13 1822

View Full Product Details